Direkt zum Inhalt
Merck

Fortfahren mit

PH010682

4-Amino-N-(4,6-dimethyl-2-pyridinyl)benzamide

AldrichCPR

Anmelden zur Ansicht der Organisations- und Vertragspreise.



SMILES string

O=C(NC1=NC(C)=CC(C)=C1)C(C=C2)=CC=C2N

InChI

1S/C14H15N3O/c1-9-7-10(2)16-13(8-9)17-14(18)11-3-5-12(15)6-4-11/h3-8H,15H2,1-2H3,(H,16,17,18)

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.


Piktogramme

Exclamation markEnvironment

Signalwort

Warning

Gefahrencodes

Hazard Classifications

Aquatic Chronic 1 - Eye Irrit. 2 - Skin Sens. 1

Lagerklasse

11 - Combustible Solids

Was ist los?

WGK 3

Flammpunkt (°F)

Not applicable

Flammpunkt (°C)

Not applicable



Hier finden Sie alle aktuellen Versionen:

Analysenzertifikate (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumente section.

Wenn Sie Hilfe benötigen, wenden Sie sich bitte an Kundensupport

Besitzen Sie dieses Produkt bereits?

In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.

Die Dokumentenbibliothek aufrufen



Questions

Reviews

No rating value

Active Filters