Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
| Größe/SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
UNSPSC Code:
12352202
NACRES:
NA.28
eCl@ss:
32160405
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltenGeneral description
48kDa
FUNCTION: SwissProt: O00141 # Protein kinase that plays an important role in cellular stress response. Activates certain potassium, sodium, and chloride channels, suggesting an involvement in the regulation of processes such as cell survival, neuronal excitability, and renal sodium excretion. Sustained high levels and activity may contribute to conditions such as hypertension and diabetic nephropathy. Mediates cell survival signals, phosphorylates and negatively regulates pro-apoptotic FOXO3A. Phosphorylates NEDD4L, which leads to its inactivation and to the subsequent activation of various channels and transporters such as ENaC, Kv1.3, or EAAT1.
SIZE: 431 amino acids; 48942 Da
SUBUNIT: Interacts with NEDD4 and NEDD4L.
SUBCELLULAR LOCATION: Cytoplasm. Nucleus. Endoplasmic reticulum. Note=Nuclear, upon phosphorylation.
TISSUE SPECIFICITY: Expressed in most tissues with highest levels in the pancreas, followed by placenta, kidney and lung.
PTM: Regulated by phosphorylation. Phosphoinositide 3-kinase (PI3- kinase) pathway promotes phosphorylation at Ser-422 which in turn increases the phosphorylation of Thr-256 by PDPK1. & Ubiquitinated by NEDD4L; which promotes proteasomal degradation. Ubiquitinated by SYVN1 at the endoplasmic reticulum; which promotes rapid proteasomal degradation and maintains a high turnover rate in resting cells.
SIMILARITY: SwissProt: O00141 ## Belongs to the protein kinase superfamily. Ser/Thr protein kinase family. & Contains 1 AGC-kinase C-terminal domain. & Contains 1 protein kinase domain.
SIZE: 431 amino acids; 48942 Da
SUBUNIT: Interacts with NEDD4 and NEDD4L.
SUBCELLULAR LOCATION: Cytoplasm. Nucleus. Endoplasmic reticulum. Note=Nuclear, upon phosphorylation.
TISSUE SPECIFICITY: Expressed in most tissues with highest levels in the pancreas, followed by placenta, kidney and lung.
PTM: Regulated by phosphorylation. Phosphoinositide 3-kinase (PI3- kinase) pathway promotes phosphorylation at Ser-422 which in turn increases the phosphorylation of Thr-256 by PDPK1. & Ubiquitinated by NEDD4L; which promotes proteasomal degradation. Ubiquitinated by SYVN1 at the endoplasmic reticulum; which promotes rapid proteasomal degradation and maintains a high turnover rate in resting cells.
SIMILARITY: SwissProt: O00141 ## Belongs to the protein kinase superfamily. Ser/Thr protein kinase family. & Contains 1 AGC-kinase C-terminal domain. & Contains 1 protein kinase domain.
Preparation Note
6 months at -70°C
Analysis Note
For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.
routinely evaluated by phosphorylation of Akt/SGK Substrate
routinely evaluated by phosphorylation of Akt/SGK Substrate
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Signalwort
Warning
Gefahrencodes
Sicherheitshinweise
Hazard Classifications
Skin Sens. 1
Lagerklasse
12 - Non Combustible Liquids
WGK
WGK 2
Flammpunkt (°F)
Not applicable
Flammpunkt (°C)
Not applicable
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?
In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.
