Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
| Größe/SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
UNSPSC Code:
12161503
NACRES:
NA.84
eCl@ss:
32161000
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltenGeneral description
OxyBlot Protein Standard is composed of five proteins (phosphorylase B, BSA, ovalbumin, carbonic anhydrase, trypsin inhibitor) containing 1-3 dinitrophenylhydrazone (DNP) residues per protein. These attached DNP moieties permit the visualization of the proteins by the detection system of the OxyBlot Protein Oxidation Detection Kit.
Application
OxyBlot Protein Standard serves as an internal control for the electrophoresis, western transfer & immunodetection steps of the OxyBlot procedure.
Preparation Note
Material Provided:
Each vial contains 150 μl of OxyBlot Protein Standard, which is sufficient for 60 minigels.
Instructions for Use:
Warm the mixture to room temperature. (Since the mixture contains SDS that may precipitate during storage, it may be necessary to heat the stock solution to 50°C to redissolve the SDS). Combine 2.5 μl of the OxyBlot Protein Standard with 20 μl of 1X Gel Loading Buffer prior to loading onto the minigel. Do not heat prior to gel loading. Perform SDS-PAGE according to standard procedures.
NOTE: Since the proteins in the standard mixture already contain DNP residues, the protein standard should not be subjected to the derivatization reaction in the OxyBlot Protein Oxidation Detection Kit protocol.
In order to maintain consistency, the 2-Mercaptoethanol must be omitted from the 1X Gel Loading Buffer if a reducing agent was not used in the preparation of the experimental samples.
Reagent Preparation:
1X Gel Loading Buffer
To make 8 ml:
Divide into 1 ml aliquots and store at -15°C to -25°C.
Protein Molecular Weight:
Phosphorylase B 97,400
Bovine serum albumin 68,000
Ovalbumin 43,000
Carbonic anhydrase 29,000
Trypsin inhibitor 21,000
Each vial contains 150 μl of OxyBlot Protein Standard, which is sufficient for 60 minigels.
Instructions for Use:
Warm the mixture to room temperature. (Since the mixture contains SDS that may precipitate during storage, it may be necessary to heat the stock solution to 50°C to redissolve the SDS). Combine 2.5 μl of the OxyBlot Protein Standard with 20 μl of 1X Gel Loading Buffer prior to loading onto the minigel. Do not heat prior to gel loading. Perform SDS-PAGE according to standard procedures.
NOTE: Since the proteins in the standard mixture already contain DNP residues, the protein standard should not be subjected to the derivatization reaction in the OxyBlot Protein Oxidation Detection Kit protocol.
In order to maintain consistency, the 2-Mercaptoethanol must be omitted from the 1X Gel Loading Buffer if a reducing agent was not used in the preparation of the experimental samples.
Reagent Preparation:
1X Gel Loading Buffer
To make 8 ml:
- 0.5 M Tris-HCl, pH 6.8
- 1.0 ml Glycerol
- 0.8 ml 14.3 M 2-Mercaptoethanol (Reagent Grade)
- 0.1 ml 10% SDS 1.6 ml
- 0.05% Bromophenol Blue 0.32 ml
- dH2O 4.18 ml
Divide into 1 ml aliquots and store at -15°C to -25°C.
Protein Molecular Weight:
Phosphorylase B 97,400
Bovine serum albumin 68,000
Ovalbumin 43,000
Carbonic anhydrase 29,000
Trypsin inhibitor 21,000
Store at -25° to -15°C upon receipt. After using for the first time, divide the remaining mixture among several tubes. Dividing the mixture into aliquots serves to protect the mixture from excessive freeze-thaw cycles.
Other Notes
Each vial contains 150 μl of OxyBlot Protein Standard, which is sufficient for 60 minigels.
OxyBlot Protein Standard serves as an internal control for the electrophoresis, western transfer and immunodetection steps of the OxyBlot procedure.
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description - | description The OxyBlot Protein Oxidation Detection Kit provides the reagents to perform the immunoblot detection of carbonyl groups introduced into proteins by oxidative reactions with ozone or oxides of nitrogen or by metal catalyzed oxidation. | description - | description clear, size 7.3 cm × 3.0 cm × 1.9 cm, pk of 5 |
| technique(s) western blot: suitable | technique(s) western blot: suitable | technique(s) western blot: suitable, immunodetection: suitable | technique(s) - |
| Quality Level 100 | Quality Level 100 | Quality Level - | Quality Level - |
| manufacturer/tradename OxyBlot | manufacturer/tradename Chemicon®, OxyBlot | manufacturer/tradename Chemicon®, Re-Blot™ | manufacturer/tradename - |
Lagerklasse
12 - Non Combustible Liquids
WGK
WGK 2
Flammpunkt (°F)
Not applicable
Flammpunkt (°C)
Not applicable
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?
In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.



