Direkt zum Inhalt
Merck

Fortfahren mit

SCC0583

3dGRO® Human Liver Organoids (MNFBQ01LI(47-49))

Normal, Non-Tumor, Age: 50 years, Female

Synonym(e):

Hepatic PDOs, Hepatocyte Organoids, Human Hepatic Organoids, Human Liver PDOs

Anmelden zur Ansicht der Organisations- und Vertragspreise.

Größe auswählen

Ansicht ändern
Größe/SKUVerfügbarkeitPreis

Über diesen Artikel

UNSPSC Code:
41106514
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten


Produktname

3dGRO® Human Liver Organoids (MNFBQ01LI(47-49)), Normal, Liver, Age: 50, Female, Mutation: None

biological source

human liver

Quality Segment

packaging

vial of ≥1500 organoids/vial vial

manufacturer/tradename

Millipore

gender

female

technique(s)

cell culture: suitable

application(s)

3D cell culture

control

liver

relevant disease(s)

none

storage temp.

-140 to -196°C

General description

Patient derived organoids (PDOs) are novel in vitro 3D cell models that have tissue-like functionality and are an alternative to traditional 2D cell lines and PDX animal models 1,2. PDOs can be derived from adult patient biopsies or resected tissues containing native LGR5+ stem cell populations and cultured within an ECM-rich substrate using specialized organoid media 3. Adult tissue derived organoids are thought to have more mature phenotypes than iPSC derived organoids and have been shown to predict patient responses to chemotherapeutics 4.
MilliporeSigma′s 3dGRO® normal liver organoid biobank is a collection of tissue-derived normal, undiseased human liver organoids. The normal liver organoid biobank contains highly characterized organoid lines from healthy donors. The 3DGRO® normal liver organoid biobank consists of Immature Liver Organoids (ILO) which can be thawed, passaged and banked back. The ILO express markers for both hepatocytes and cholangiocytes, and when needed, they may be differentiated into Mature Liver Organoids (MLO), which are terminally differentiated.

Patient Specific Details
Organ Type: Liver
Patient Age: 50 years
Patient Species: Human
Sex: Female
Race: Asian Pacific
Disease: Normal
Tumor Source: Non-Tumor
Model ID: MNFBQ01LI47

Application

Specifications
Viability: >1500 organoids/vial
Organoid Growth: The organoids can be cultured for at least 5 passages after initial thawing without significantly affecting the cell marker expression and functionality.
Species Verification: Species has been verified and confirmed to be negative for inter-species contamination by a Contamination Clear panel by Charles River Animal Diagnostic Services.
  • Infectious Disease Testing: Cells tested negative for infectious disease by a PCR panel. Human lines negative for HPV-16, HPV-18, Hepatitis A, B, C, and HIV-1 & 2 viruses.
  • Microbial Contamination: None detected.
  • Mycoplasma Contamination: Negative
  • STR Profile: Pass
  • Characterization: Immunocytochemistry

Preparation Note

These organoids should be stored in liquid nitrogen.

Other Notes

Yang H, Sun L, Liu M, Mao Y. 2018. Patient-derived organoids: a promising model for personalized cancer treatment Gastroenterol Rep (Oxf). 6(4):243–245.Margit Bleijs M, van de Wetering M, Clevers H, Drost J. 2019. Xenograft and organoid model systems in cancer research. EMBO J. 38(15):e101654.Broutier L, Andersson-Rolf A, Hindley CJ, Boj SF, Clevers H, Koo B, Huch M. 2016. Culture and establishment of self-renewing human and mouse adult liver and pancreas 3D organoids and their genetic manipulation. Nat. Protoc. 11(9):1724-43.Harrison SP, Baumgarten SP, Verma R, Oleg Lunov O, Dejneka A, Sullivan GJ. 2021. Liver Organoids: Recent Developments, Limitations and Potential. Front. Med. (Lausanne). 8:574047

Legal Information

3dGRO® is a trademark of Merck KGaA, Darmstadt, Germany.
3dGRO® organoids were derived utilizing HUB Organoid Technology.

Subject to local law, this product is intended to be sold for internal in vitro research use only subject to terms and conditions found here: www.sigmaaldrich.com/pdouse. This product may not be re-engineered or copied; used to make derivatives or modifications; used to develop, test, or manufacture a commercial product; used as a component in a commercial product; resold or licensed; used in any clinical applications or trials; or used in humans. A license or limited commercial use agreement is required for use by any for-profit entity, use in services, and use in sponsored academic research. For information regarding any such use, please contact organoids@milliporesigma.com.
3dGRO is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Ähnliche Artikel vergleichen

Vollständigen Vergleich anzeigen

Unterschiede anzeigen

1 of 1

Dieser Artikel
SCC585SCC589SCC587
description

Normal, Liver, Age: 50, Female, Mutation: None

description

Normal, Liver, Age: 57, Female, Mutation: None

description

Normal, Liver, Age: 51, Female, Mutation: None

description

Normal, Liver, Age: 60, Male, Mutation: None

application(s)

3D cell culture

application(s)

3D cell culture

application(s)

3D cell culture

application(s)

3D cell culture

technique(s)

cell culture: suitable

technique(s)

cell culture: suitable

technique(s)

cell culture: suitable

technique(s)

cell culture: suitable

relevant disease(s)

none

relevant disease(s)

(normal)

relevant disease(s)

(normal)

relevant disease(s)

(normal)

biological source

human liver

biological source

human liver

biological source

human liver

biological source

human liver

storage temp.

-140 to -196°C

storage temp.

-140 to -196°C

storage temp.

-140 to -196°C

storage temp.

-140 to -196°C

gender

female

gender

female

gender

female

gender

male


Lagerklasse

10 - Combustible liquids

Flammpunkt (°F)

Not applicable

Flammpunkt (°C)

Not applicable


Zulassungslistungen

Zulassungslistungen werden hauptsächlich für chemische Produkte erstellt. Für nicht-chemische Produkte können hier nur begrenzte Angaben gemacht werden. Kein Eintrag bedeutet, dass keine der Komponenten gelistet ist. Es liegt in der Verantwortung des Benutzers, die sichere und legale Verwendung des Produkts zu gewährleisten.

64-17-5

CAS No.


Hier finden Sie alle aktuellen Versionen:

Analysenzertifikate (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumente section.

Wenn Sie Hilfe benötigen, wenden Sie sich bitte an Kundensupport

Besitzen Sie dieses Produkt bereits?

In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.

Die Dokumentenbibliothek aufrufen