Direkt zum Inhalt
Merck

Fortfahren mit

SCC581

3dGRO® Human Liver Organoids MNFBQ01LI (38-43)

Normal, Liver, Age: 60, Female, Mutation: None

Synonym(e):

Hepatic PDOs, Hepatocyte Organoids, Human Hepatic Organoids, Human Liver PDOs

Anmelden zur Ansicht der Organisations- und Vertragspreise.

Größe auswählen

Ansicht ändern
Größe/SKUVerfügbarkeitPreis

Über diesen Artikel

UNSPSC Code:
41106514
NACRES:
NA.61
eCl@ss:
32160703
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten


Quality Segment

biological source

human liver

packaging

vial of ≥1500 vial (viable cells per vial)

manufacturer/tradename

Millipore

gender

female

technique(s)

cell culture: suitable

application(s)

3D cell culture

control

liver

relevant disease(s)

(normal)

shipped in

liquid nitrogen

storage temp.

−196°C

General description

Patient derived organoids (PDO) are tissue derived, novel in vitro 3D cell models that recapitulate some key features of the respective organ, thus representing a more in vivo-like alternative to traditional 2D cell cultures. In addition, PDOs offer possible means to reduce animal model use. PDO maybe derived from resections or biopsy of adult tissue and are phenotypically and genetically stable in culture. PDO are an ideal platform for drug screening, disease modeling and tumor immunity studies, and may also be modified by gene editing.

The 3dGRO® human liver organoid biobank is a collection of tissue-derived normal non-diseased human liver organoids from numerous patients. The biobank contains liver PDOs that are highly viable and express high levels of albumin, HNF4-a, FOXA2, SOX17, CK7, CK19, E-Cadherin, PCK2 along with important cytochrome P450 (CYP) enzymes activities (Cyp3A4, Cyp2C9, Cyp2B6, Cyp2D6, Cyp1A2).

Cell Line Characteristic

  • Sex: Female
  • Age: 60 Years
  • Race: Asian (Indian)
  • Organ: Liver
  • Disease: Normal
  • Patient History: None

Application

  • Viability: >1500 viable organoids/vial
  • Organoid Growth: Pass
  • Cells are tested negative for infectious diseases by a Human Essential CLEAR panel by Charles River Animal Diagnostic Services.
  • Mycoplasma Contamination: Negative
  • STR Profile: Pass

Features and Benefits

Cryopreserved patient derived human liver organoids (PDOs)

Other Notes

Store in liquid nitrogen. The organoids can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality.

Legal Information

3dGRO® is a trademark of Merck KGaA, Darmstadt, Germany.
3dGRO® organoids were derived utilizing HUB Organoid Technology.

Subject to local law, this product is intended to be sold for internal in vitro research use only subject to terms and conditions found here: www.sigmaaldrich.com/pdouse. This product may not be re-engineered or copied; used to make derivatives or modifications; used to develop, test, or manufacture a commercial product; used as a component in a commercial product; resold or licensed; used in any clinical applications or trials; or used in humans. A license or limited commercial use agreement is required for use by any for-profit entity, use in services, and use in sponsored academic research. For information regarding any such use, please contact organoids@milliporesigma.com.
3dGRO is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

This product is regulated in France when intended for scientific purposes, including import, export, and conservation activities (Articles L.1211-1, paragraph 2, and L.1243-4 of the French Public Health Code). The purchaser (i.e. end user) is required to obtain an import authorization from the France Ministry responsible for Research (as referenced in Article L.1245-5-1 II of the French Public Health Code) or a conservation authorization from the same ministry (as referenced in Article L.1243-4 of the French Public Health Code). By placing an order for this product, you confirm that you have obtained the appropriate import or conservation authorization for the product. For more information regarding the use of human biological samples for research purposes in France, please contact the relevant French authorities.
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Ähnliche Artikel vergleichen

Vollständigen Vergleich anzeigen

Unterschiede anzeigen

1 of 1

Dieser Artikel
SCC589SCC0583SCC588
description

Normal, Liver, Age: 60, Female, Mutation: None

description

Normal, Liver, Age: 51, Female, Mutation: None

description

Normal, Liver, Age: 50, Female, Mutation: None

description

Normal, Liver, Age: 65, Female, Mutation: None

application(s)

3D cell culture

application(s)

3D cell culture

application(s)

3D cell culture

application(s)

3D cell culture

relevant disease(s)

(normal)

relevant disease(s)

(normal)

relevant disease(s)

none

relevant disease(s)

(normal)

technique(s)

cell culture: suitable

technique(s)

cell culture: suitable

technique(s)

cell culture: suitable

technique(s)

cell culture: suitable

biological source

human liver

biological source

human liver

biological source

human liver

biological source

human liver

storage temp.

−196°C

storage temp.

-140 to -196°C

storage temp.

-140 to -196°C

storage temp.

−196°C

gender

female

gender

female

gender

female

gender

female


Lagerklasse

10 - Combustible liquids

Flammpunkt (°F)

Not applicable

Flammpunkt (°C)

Not applicable



Hier finden Sie alle aktuellen Versionen:

Analysenzertifikate (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumente section.

Wenn Sie Hilfe benötigen, wenden Sie sich bitte an Kundensupport

Besitzen Sie dieses Produkt bereits?

In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.

Die Dokumentenbibliothek aufrufen