Direkt zum Inhalt
Merck

Fortfahren mit

SCC230

YUMM1.G1 Mouse Melanoma Cell Line

Synonym(e):

YUMM1.G1 Mouse Melanoma Cell Line, Yale University Mouse Melanoma 1.G1A cell line, Yale University Mouse Melanoma cell line

Anmelden zur Ansicht der Organisations- und Vertragspreise.

Größe auswählen

Ansicht ändern
Größe/SKUVerfügbarkeitPreis

Über diesen Artikel

UNSPSC Code:
41106514
NACRES:
NA.51
Biological source:
mouse
Manufacturer/tradename:
Millipore
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten


biological source

mouse

Quality Segment

packaging

vial of ≥1X10⁶ vial (viable cells per vial)

manufacturer/tradename

Millipore

technique(s)

cell culture | mammalian: suitable

shipped in

liquid nitrogen

storage temp.

−196°C

General description

As immune-based therapies in cancer models become more popular and applicable, a further need for immune-competent models has been presented. A wide variety of genetically engineered mouse models have been created to express vital melanoma genotypes. The variety in these models have allowed the study of genetic alterations that are relevant to human melanoma biology. While it is quite useful to have mouse models with genetically defined human melanoma characteristics, itis limited by complex mouse colony maintenance. The YUMM1.G1 cell line was produced to be more experimentally relevant. The cell line was derived by backcrossing important alleles into C57BL/6J mice. The mice were interbred to produce human-relevant genetically defined mouse models that could then be used to generate cell lines. Melanomas were induced by applying 4-hydroxytamoxifen to induce Cre-lox allelere combination. Cells were then mechanically and enzymatically dissociated from the removed tumor. The melanoma lines generated from this procedure retained important human melanoma characteristics useful for studying disease progression.The YUMM1.G1 cell line is derived from a Mc1re/e female mouse model driven by Braf activation, Pten inactivation, and Cdkn2ainactivation. The cell line is positive for the common melanoma markers, MelanA and PMEL. The cells are syngeneic withC57BL/6J mice and are very tumorigenic in vivo. References:1. Meeth K et al. (2016) The YUMM lines: a series of congenic mouse melanoma cell lines with defined genetic alterations. Pigment Cell Melanoma Res 29(5): 590-597.

Application

  • Each vial contains > 1X106 viable cells.
  • Cells are tested negative for infectious diseases by a Mouse Essential CLEAR panel by Charles River Animal Diagnostic Services.
  • Cells are verified to be of mouse origin and negative for inter-species contamination from rat, chinese hamster, Golden Syrian hamster, human and non-human primate (NHP) as assessed by a Contamination CLEAR panel by Charles River Animal Diagnostic Services.
  • Cells are negative for mycoplasma contamination.

Features and Benefits

YUMM1.G1 cell line is derived from a Mc1re/e female mouse model driven by Braf activation, Pten inactivation, and Cdkn2a inactivation. The cell line is positive for the common melanoma markers, MelanA and PMEL.

Preparation Note

YUMM1.G1A cells should be stored in liquid nitrogen until use. The cells can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality.

Analysis Note

This product is intended for sale and sold solely to academic institutions for internal academic research use per the terms of the “Academic Use Agreement” as detailed in the product documentation. For information regarding any other use, please contact licensing@milliporesigma.com.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Ähnliche Artikel vergleichen

Vollständigen Vergleich anzeigen

Unterschiede anzeigen

1 of 1

Dieser Artikel
SCC226SCC237SCC231
description

-

description

-

description

-

description

-

biological source

mouse

biological source

mouse

biological source

mouse

biological source

mouse

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

storage temp.

−196°C

storage temp.

−196°C

storage temp.

−196°C

storage temp.

−196°C

shipped in

liquid nitrogen

shipped in

liquid nitrogen

shipped in

liquid nitrogen

shipped in

liquid nitrogen

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

manufacturer/tradename

Millipore

manufacturer/tradename

Millipore

manufacturer/tradename

Millipore

manufacturer/tradename

Millipore


Lagerklasse

12 - Non Combustible Liquids



Hier finden Sie alle aktuellen Versionen:

Analysenzertifikate (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumente section.

Wenn Sie Hilfe benötigen, wenden Sie sich bitte an Kundensupport

Besitzen Sie dieses Produkt bereits?

In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.

Die Dokumentenbibliothek aufrufen