Direkt zum Inhalt
Merck

Fortfahren mit

33673

6-Chlorbenzoxazol-2(3H)-on

PESTANAL®, analytical standard

Anmelden zur Ansicht der Organisations- und Vertragspreise.

Über diesen Artikel

Empirische Formel (Hill-System):
C7H4ClNO2
CAS-Nummer:
Molekulargewicht:
169.57
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:


grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, chromatography: suitable, gas chromatography (GC): suitable

application(s)

environmental

format

neat

SMILES string

Clc1ccc2NC(=O)Oc2c1

InChI

1S/C7H4ClNO2/c8-4-1-2-5-6(3-4)11-7(10)9-5/h1-3H,(H,9,10)

InChI key

MATCZHXABVLZIE-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany


Piktogramme

Exclamation mark

Signalwort

Warning

Gefahrencodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Zielorgane

Respiratory system

Lagerklasse

11 - Combustible Solids

Was ist los?

WGK 3

Flammpunkt (°F)

Not applicable

Flammpunkt (°C)

Not applicable

PSA (Persönliche Schutzausrüstung)

dust mask type N95 (US), Eyeshields, Gloves



Hier finden Sie alle aktuellen Versionen:

Analysenzertifikate (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumente section.

Wenn Sie Hilfe benötigen, wenden Sie sich bitte an Kundensupport

Besitzen Sie dieses Produkt bereits?

In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.

Die Dokumentenbibliothek aufrufen



Questions

Reviews

No rating value

Active Filters