Direkt zum Inhalt
Merck

Fortfahren mit

ORMS-6

Water Certified Reference Material for Mercury

certified reference material

Synonym(e):

reference material

Anmelden zur Ansicht der Organisations- und Vertragspreise.

Größe auswählen

Ansicht ändern
Größe/SKUVerfügbarkeitPreis

Über diesen Artikel

UNSPSC Code:
41116107
eCl@ss:
25140904
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten


grade

certified reference material

agency

(National Research Council of Canada (NRC))

manufacturer/tradename

National Research Council of Canada (NRC)

application(s)

food and beverages

format

(Calibration solution)

General description

ORMS-6 is a water spiked with inorganic mercury Certified Reference Material (CRM) from the National Research Council Canada (NRC). The material is packaged in a 50 mL glass ampoule stabilized with 0.5% BrCl. https://doi.org/10.4224/crm.2025.orms-6

Features and Benefits

NRC Canada′s certified reference materials (CRMs) are traceable to the International System of Units, and conform to the requirements of ISO/IEC 17025 and ISO 17034.For specific values, as well as additional information regarding stability and storage information, please view the Certificate of Analysis.With NRC Canada′s CRMs, you gain access to products backed by extensive research and expertise, fostering trust and consistency in your results. Elevate your laboratory′s accuracy and credibility with NRC Canada′s certified reference materials.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Ähnliche Artikel vergleichen

Vollständigen Vergleich anzeigen

Unterschiede anzeigen

1 of 1

Dieser Artikel
CRMAZA6BPAN1BOVM1
description

certified reference material

description

certified reference material

description

certified reference material

description

certified reference material

application(s)

food and beverages

application(s)

food and beverages

application(s)

cosmetics
food and beverages

application(s)

cosmetics
food and beverages

format

(Calibration solution)

format

(Calibration solution)

format

neat

format

matrix material

agency

(National Research Council of Canada (NRC))

agency

(National Research Council of Canada (NRC))

agency

-

agency

-

grade

certified reference material

grade

certified reference material

grade

certified reference material

grade

certified reference material

manufacturer/tradename

National Research Council of Canada (NRC)

manufacturer/tradename

National Research Council of Canada (NRC)

manufacturer/tradename

National Research Council of Canada (NRC)

manufacturer/tradename

National Research Council of Canada (NRC)


Lagerklasse

12 - Non Combustible Liquids

Flammpunkt (°F)

Not applicable

Flammpunkt (°C)

Not applicable



Hier finden Sie alle aktuellen Versionen:

Analysenzertifikate (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumente section.

Wenn Sie Hilfe benötigen, wenden Sie sich bitte an Kundensupport

Besitzen Sie dieses Produkt bereits?

In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.

Die Dokumentenbibliothek aufrufen