Direkt zum Inhalt
Merck

Fortfahren mit

SLEW-4

Estuarine water certified reference material for trace metals

certified reference material

Synonym(e):

Estuarine water certified reference material for trace metals

Anmelden zur Ansicht der Organisations- und Vertragspreise.

Größe auswählen

Ansicht ändern
Größe/SKUVerfügbarkeitPreis

Über diesen Artikel

NACRES:
NA.24
UNSPSC Code:
41116107
eCl@ss:
25140904
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten


grade

certified reference material

agency

traceable to (National Research Council of Canada (NRC))

manufacturer/tradename

National Research Council of Canada (NRC)

application(s)

food and beverages

format

matrix material

General description

NRC Canada’s certified reference materials (CRMs) are traceable to the International System of Units, and conform to the requirements of ISO/IEC 17025 and ISO 17034.For specific values, as well as additional information regarding stability and storage information, please view the Certificate of Analysis.With NRC Canada’s CRMs, you gain access to products backed by extensive research and expertise, fostering trust and consistency in your results. Elevate your laboratory’s accuracy and credibility with NRC Canada’s certified reference materials.
SLEW-4 is an estuarine water Certified Reference Material (CRM) from the National Research Council Canada (NRC) with information on trace element content. A unit of SLEW-4 consists of approximately 200 mL of estuarine water. This material is intended for use in the method development, validation, and quality control for the analysis of trace and matrix constituents in seawater matrices. For more information, please refer to the Certificate of Analysis or Safety Data Sheet.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

Tested for the analytes listed below. See certificate for values and more details


Trace Elements: Arsenic (As), Boron (B), Cadmium (Cd), Chromium (Cr), Cobalt (Co), Copper (Cu), Iron (Fe), Lead (Pb), Manganese (Mn), Molybdenum (Mo), Nickel (Ni), Uranium (U), Vanadium (V), Zinc (Zn)


Lagerklasse

10 - Combustible liquids



Hier finden Sie alle aktuellen Versionen:

Analysenzertifikate (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumente section.

Wenn Sie Hilfe benötigen, wenden Sie sich bitte an Kundensupport

Besitzen Sie dieses Produkt bereits?

In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.

Die Dokumentenbibliothek aufrufen