Fortfahren mit
Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
biological source
non-animal source
Quality Segment
agency
EP, JP, USP
product line
EMPROVE® EXPERT
assay
≥98.5%
form
crystalline powder
technique(s)
cell culture | mammalian: suitable
impurities
endotoxin, heavy metals, residual solvents tested
mp
228 °C (dec.) (lit.)
solubility
H2O: 50 mg/mL
application(s)
pharmaceutical (small molecule)
foreign activity
cytotoxicity, tested
SMILES string
OC(=O)[C@@H]1CCCN1
InChI
1S/C5H9NO2/c7-5(8)4-2-1-3-6-4/h4,6H,1-3H2,(H,7,8)/t4-/m0/s1
InChI key
ONIBWKKTOPOVIA-BYPYZUCNSA-N
General description
M-Clarity Program
As part of our Emprove® Program, our raw materials are offered with Emprove® Dossiers which provide comprehensive, up-to-date documentation to help you navigate regulatory challenges, manage risks, and improve your manufacturing processes.
Application
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description EMPROVE® EXPERT, EP, JP, USP | description certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland | description - | description ReagentPlus®, ≥99% (HPLC) |
| mp 228 °C (dec.) (lit.) | mp 228 °C (dec.) (lit.) | mp 228 °C (dec.) (lit.) | mp 228 °C (dec.) (lit.) |
| technique(s) cell culture | mammalian: suitable | technique(s) HPLC: suitable, gas chromatography (GC): suitable | technique(s) cell culture | mammalian: suitable | technique(s) - |
| form crystalline powder | form - | form crystalline powder | form powder |
| foreign activity cytotoxicity, tested | foreign activity - | foreign activity cytotoxicity, tested | foreign activity - |
| impurities endotoxin, heavy metals, residual solvents tested | impurities - | impurities endotoxin, heavy metals, residual solvents, tested | impurities L-Hydroxyproline, free |
| solubility H2O: 50 mg/mL | solubility - | solubility H2O: 50 mg/mL | solubility - |
Still not finding the right product?
Explore all of our products under L-Prolin
Lagerklasse
11 - Combustible Solids
Was ist los?
WGK 1
Flammpunkt (°F)
Not applicable
Flammpunkt (°C)
Not applicable
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?
In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.
