Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|---|---|
2 ea | Warenkorb auf Verfügbarkeit prüfen | € 60,70 |
Über diesen Artikel
€ 60,70
material
polypropylene screw closure, retained silicone gasket
packaging
pack of 2 ea
manufacturer/tradename
Nalgene DS3132-0063
diam.
63 mm
General description
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description diam. 63 mm, for use with 500 mL and 1 L tubes, pack of 2 ea | description diam. 20 mm, for use with 28 and 30 mL tubes, pack of 10 ea | description diam. 24 mm, for use with 50 mL tubes, pack of 10 ea | description polypropylene cap, diam. × thread 22 mm × 20-415 mm, case of 2000 ea |
| manufacturer/tradename Nalgene DS3132-0063 | manufacturer/tradename Nalgene DS3132-0020 | manufacturer/tradename Nalgene DS3132-0024 | manufacturer/tradename Nalgene 362150-5200 |
| material polypropylene screw closure, retained silicone gasket | material polypropylene screw closure, retained silicone gasket | material polypropylene screw closure, retained silicone gasket | material polypropylene cap, red cap |
| packaging pack of 2 ea | packaging pack of 10 ea | packaging pack of 10 ea | packaging case of 2000 ea |
| diam. 63 mm | diam. 20 mm | diam. 24 mm | diam. - |
Still not finding the right product?
Explore all of our products under Nalgene® sealing caps
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?