Fortfahren mit
Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
Quality Segment
sterility
0.2 μm filtered
product line
BioReagent
form
liquid
shelf life
2 yr (retest)
technique(s)
cell culture | mammalian: suitable
shipped in
ambient
storage temp.
2-8°C
General description
Application
Biochem/physiol Actions
Antimicrobial Spectrum: G418 selects for cells stably transfected with an iNOS promoter construct and neomycin resistance gene and shows activity against protozoa and helminthes.
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description 50 mg/mL in H2O, 0.2 μm filtered, BioReagent, suitable for cell culture | description 1 mg/mL in deionized water, 0.2 μm filtered, BioReagent, suitable for cell culture | description 50 mg/mL in H2O, 0.1 μm filtered, BioReagent, suitable for cell culture | description solution, =98% (TLC), suitable for transfection |
| technique(s) cell culture | mammalian: suitable | technique(s) cell culture | mammalian: suitable | technique(s) cell culture | mammalian: suitable | technique(s) transfection: suitable |
| Quality Level 300 | Quality Level 300 | Quality Level 200 | Quality Level - |
| product line BioReagent | product line BioReagent | product line BioReagent | product line - |
| form liquid | form liquid | form solution | form solution |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C |
| shipped in ambient | shipped in ambient | shipped in - | shipped in - |
Signalwort
Danger
Gefahrencodes
Sicherheitshinweise
Hazard Classifications
Resp. Sens. 1 - Skin Sens. 1
Lagerklasse
12 - Non Combustible Liquids
WGK
WGK 3
Flammpunkt (°F)
Not applicable
Flammpunkt (°C)
Not applicable
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?
In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.



