Fortfahren mit
GILFA10012
Gilson™ PIPETMAN® L Multichannel Pipette
P12x200L, 12-channel, volume 20-200 μL
Synonym(e):
manual pipettor, mechanical pipette, micropipette, pipette
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
Größe/SKUVerfügbarkeitPreis
Über diesen Artikel
UNSPSC Code:
41121500
eCl@ss:
32039090
Volume:
20-200 μL
Packaging:
pack of 1 ea
Feature:
adjustable, electronic: no, 12-channel
Manufacturer/tradename:
Gilson FA10012
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten
material
yellow (color code)
description
calibrated: yes
feature
adjustable, electronic: no, 12-channel
packaging
pack of 1 ea
manufacturer/tradename
Gilson FA10012
volume
20-200 μL
compatibility
Gilson PIPETMAN DIAMOND Tips D200, D300, DF200ST, DF300ST
General description
PIPETMAN® L multichannel air-displacement pipettes cover a volume range from 0.5 μL to 1200 μL, including four models equipped with V-rings (VR) at the bottom of the tip holders for a proper fit with most tip brands.
Application
Biology, Biochemistry, Immunoassays, Cell culture, Analytical chemistry
Features and Benefits
  • Convenient Use - Lightweight and balanced, PIPETMAN® L combines an ergonomic design with minimized pipetting forces, giving you the ability to pipette longer without fatigue.
  • Right-handed or left-handed, no problem - Adjust the location of the tip ejector to the most comfortable position for you. PIPETMAN® L single channel models are equipped with a stainless-steel tip ejector. Should your application require to pipette corrosive liquids, plastic tip ejectors are available as accessories. P5000L and P10mLL models are equipped with a plastic tip ejector to avoid direct contact with the tip and provide better compatibility with chemicals.
  • Secure Pipetting - Reduce pipetting errors and avoid accidental volume changes during pipetting cycles with a true volume-locking mechanism for improved accuracy and precision.
  • PIPETMAN® L models are all fully autoclavable without disassembly - maximum convenience and protection from cross-contamination.
  • GLP Compliance - PIPETMAN® L models feature a DataMatrix 2D code for easy traceability. Use a 2D code scanner to transfer the code automatically into any traceability or asset management system. Also, the pipette serial number is engraved on the body of the pipette for unique identification.
Legal Information
Gilson is a trademark of Gilson, Inc.
Pipetman is a registered trademark of Gilson, Inc.
Ähnliche Artikel vergleichen
Vollständigen Vergleich anzeigen
Unterschiede anzeigen
1 of 1
Dieser Artikel
GILFA10011GILFA10036GILFA10016
description
P12x200L, 12-channel, volume 20-200 μL
description
P8x200L, 8-channel, volume 20-200 μL
description
P12x200L, 12-channel, volume 20-200 μL, with V-Rings
description
P12x300L, 12-channel
feature
adjustable, electronic: no, 12-channel
feature
adjustable, 8-channel, electronic: no
feature
electronic: no, adjustable, with V-Rings, 12-channel
feature
adjustable, electronic: no, 12-channel
manufacturer/tradename
Gilson FA10012
manufacturer/tradename
Gilson FA10011
manufacturer/tradename
Gilson FA10036
manufacturer/tradename
Gilson FA10016
volume
20-200 μL
volume
20-200 μL
volume
20-200 μL
volume
20-300 μL
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
pack of 1 ea
material
yellow (color code)
material
yellow (color code)
material
-
material
-
compatibility
Gilson PIPETMAN DIAMOND Tips D200, D300, DF200ST, DF300ST
compatibility
Gilson PIPETMAN DIAMOND Tips D200, D300, DF200ST, DF300ST
compatibility
Gilson PIPETMAN DIAMOND Tips D200, D300, DF200ST, DF300ST
compatibility
Gilson PIPETMAN DIAMOND Tips D200, D300, DF200ST, DF300ST
Still not finding the right product?
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)