Direkt zum Inhalt
Merck

Fortfahren mit

GILFA10036

Gilson™ PIPETMAN® L Multichannel Pipette

P12x200L, 12-channel, volume 20-200 μL, with V-Rings

Synonym(e):

manual pipettor, mechanical pipette, micropipette, pipette

Anmelden zur Ansicht der Organisations- und Vertragspreise.

Größe auswählen

Ansicht ändern
Größe/SKUVerfügbarkeitPreis

Über diesen Artikel

UNSPSC Code:
41121500
eCl@ss:
32070102
Volume:
20-200 μL
Packaging:
pack of 1 ea
Feature:
adjustable, electronic: no, 12-channel, with V-Rings
Manufacturer/tradename:
Gilson FA10036
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten


material

yellow (color code)

description

calibrated: yes

feature

adjustable, electronic: no, 12-channel, with V-Rings

packaging

pack of 1 ea

manufacturer/tradename

Gilson FA10036

volume

20-200 μL

compatibility

Gilson PIPETMAN DIAMOND Tips D200, D300, DF200ST, DF300ST

General description

PIPETMAN® L multichannel air-displacement pipettes cover a volume range from 0.5 μL to 1200 μL, including four models equipped with V-rings (VR) at the bottom of the tip holders for a proper fit with most tip brands.

Application

Biology, Biochemistry, Immunoassays, Cell culture, Analytical chemistry

Features and Benefits

  • Convenient Use - Lightweight and balanced, PIPETMAN® L combines an ergonomic design with minimized pipetting forces, giving you the ability to pipette longer without fatigue.
  • Right-handed or left-handed, no problem - Adjust the location of the tip ejector to the most comfortable position for you. PIPETMAN® L single channel models are equipped with a stainless-steel tip ejector. Should your application require to pipette corrosive liquids, plastic tip ejectors are available as accessories. P5000L and P10mLL models are equipped with a plastic tip ejector to avoid direct contact with the tip and provide better compatibility with chemicals.
  • Secure Pipetting - Reduce pipetting errors and avoid accidental volume changes during pipetting cycles with a true volume-locking mechanism for improved accuracy and precision.
  • PIPETMAN® L models are all fully autoclavable without disassembly - maximum convenience and protection from cross-contamination.
  • GLP Compliance - PIPETMAN® L models feature a DataMatrix 2D code for easy traceability. Use a 2D code scanner to transfer the code automatically into any traceability or asset management system. Also, the pipette serial number is engraved on the body of the pipette for unique identification.

Legal Information

Gilson is a trademark of Gilson, Inc.
Pipetman is a registered trademark of Gilson, Inc.

Ähnliche Artikel vergleichen

Vollständigen Vergleich anzeigen

Unterschiede anzeigen

1 of 1

Dieser Artikel
GILFA10035GILFA10012GILFA10037
description

P12x200L, 12-channel, volume 20-200 μL, with V-Rings

description

P8x200L, 8-channel, volume 20-200 μL, with V-Rings

description

P12x200L, 12-channel, volume 20-200 μL

description

P8x300L, 8-channel, volume 20-300 μL, with V-Rings

manufacturer/tradename

Gilson FA10036

manufacturer/tradename

Gilson FA10035

manufacturer/tradename

Gilson FA10012

manufacturer/tradename

Gilson FA10037

feature

electronic: no, adjustable, with V-Rings, 12-channel

feature

8-channel, with V-Rings, adjustable, electronic: no

feature

adjustable, electronic: no, 12-channel

feature

with V-Rings, adjustable, 8-channel, electronic: no

volume

20-200 μL

volume

20-200 μL

volume

20-200 μL

volume

20-300 μL

packaging

pack of 1 ea

packaging

pack of 1 ea

packaging

pack of 1 ea

packaging

pack of 1 ea

description

calibrated: yes

description

calibrated: yes

description

calibrated: yes

description

-

compatibility

Gilson PIPETMAN DIAMOND Tips D200, D300, DF200ST, DF300ST

compatibility

Gilson PIPETMAN DIAMOND Tips D200, D300, DF200ST, DF300ST

compatibility

Gilson PIPETMAN DIAMOND Tips D200, D300, DF200ST, DF300ST

compatibility

Gilson PIPETMAN DIAMOND Tips D200, D300, DF200ST, DF300ST


Still not finding the right product?




Hier finden Sie alle aktuellen Versionen:

Analysenzertifikate (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumente section.

Wenn Sie Hilfe benötigen, wenden Sie sich bitte an Kundensupport

Besitzen Sie dieses Produkt bereits?

In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.

Die Dokumentenbibliothek aufrufen