Direkt zum Inhalt
Merck

Fortfahren mit

MIR660

TransIT®-Lenti Transfection Reagent

Ideal transfection reagent for recombinant lentivirus production

Synonym(e):

Lentivirus production, MIR 660

Anmelden zur Ansicht der Organisations- und Vertragspreise.

Größe auswählen

Ansicht ändern
Größe/SKUVerfügbarkeitPreis

Über diesen Artikel

UNSPSC Code:
41106502
Form:
liquid
Grade:
Molecular Biology
Technique(s):
transfection: suitable
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten


Produktname

TransIT®-Lenti, Ideal transfection reagent for recombinant lentivirus production

grade

Molecular Biology

form

liquid

manufacturer/tradename

Mirus Bio

technique(s)

transfection: suitable

shipped in

wet ice

storage temp.

−20°C

General description

Lentivirus is an enveloped, single-stranded RNA virus from the Retroviridae family capable of infecting both dividing and non-dividing cells. Combined with an efficient host-genome integration mechanism, this capability has led to recombinant lentivirus becoming a central gene delivery tool for robust and stable transgene expression in target cells. However, the lentivirus platform is often limited by insufficient viral titers requiring concentration before use. To address this limitation, Mirus Bio developed the TransIT® Lentivirus System for enhanced delivery and expression of the essential gag, pol, rev and VSV-G genes required for higher-titer lentivirus production in adherent or suspension HEK 293 derived cell types.

Features and Benefits

TransIT®-Lenti Transfection Reagent is designed to enhance delivery of packaging and transfer vectors to adherent HEK 293T and suspension 293-F cell types to increase recombinant lentivirus production.
  • High Performance – Provide higher functional titers
  • Simple Protocol – No media change required, single harvest
  • Animal Origin Free – Regulatory friendly

Other Notes

Animal origin free.

Suitable for lentivirus transfection.

Legal Information

TransIT is a registered trademark of Mirus Bio

Ähnliche Artikel vergleichen

Vollständigen Vergleich anzeigen

Unterschiede anzeigen

1 of 1

Dieser Artikel
MIR230MIR270MIR57
TransIT®-Lenti Ideal transfection reagent for recombinant lentivirus production

MIR660

TransIT®-Lenti

TransIT®-LT1 A low toxicity, broad spectrum DNA transfection reagent

MIR230

TransIT®-LT1

TransIT®-293 A high efficiency, low toxicity, DNA transfection reagent optimized for 293 cells

MIR270

TransIT®-293

TransIT®-PRO High, Reproducible Protein Yield in Suspension CHO and 293 Cells

MIR57

TransIT®-PRO

description

Ideal transfection reagent for recombinant lentivirus production

description

A low toxicity, broad spectrum DNA transfection reagent

description

A high efficiency, low toxicity, DNA transfection reagent optimized for 293 cells

description

High, Reproducible Protein Yield in Suspension CHO and 293 Cells

technique(s)

transfection: suitable

technique(s)

transfection: suitable

technique(s)

transfection: suitable

technique(s)

transfection: suitable

form

liquid

form

liquid

form

liquid

form

liquid

grade

Molecular Biology

grade

Molecular Biology

grade

Molecular Biology

grade

Molecular Biology

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

manufacturer/tradename

Mirus Bio

manufacturer/tradename

Mirus Bio

manufacturer/tradename

Mirus Bio

manufacturer/tradename

Mirus Bio

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice


Piktogramme

FlameCorrosion

Signalwort

Danger

Gefahrencodes

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1

Lagerklasse

3 - Flammable liquids

Flammpunkt (°F)

66.2 °F

Flammpunkt (°C)

19 °C


Zulassungslistungen

Zulassungslistungen werden hauptsächlich für chemische Produkte erstellt. Für nicht-chemische Produkte können hier nur begrenzte Angaben gemacht werden. Kein Eintrag bedeutet, dass keine der Komponenten gelistet ist. Es liegt in der Verantwortung des Benutzers, die sichere und legale Verwendung des Produkts zu gewährleisten.

64-17-5

CAS No.


Hier finden Sie alle aktuellen Versionen:

Analysenzertifikate (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumente section.

Wenn Sie Hilfe benötigen, wenden Sie sich bitte an Kundensupport

Besitzen Sie dieses Produkt bereits?

In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.

Die Dokumentenbibliothek aufrufen