Direkt zum Inhalt
Merck

Fortfahren mit

P5442

Active Recombinant Human Phosphoglucose Isomerase

Lyophilized powder, ≥ 100U/mg protein

Synonym(e):

D-Glucose-6-phosphate ketol-isomerase, PGI, Phosphosaccharomutase, GPI

Anmelden zur Ansicht der Organisations- und Vertragspreise.

Größe auswählen

Ansicht ändern
Größe/SKUVerfügbarkeitPreis

Über diesen Artikel

UNSPSC Code:
12352204
eCl@ss:
32160701
Form:
lyophilized powder
Assay:
≥90% (SDS-PAGE)
Biological source:
human
Mol wt:
~63 kDa
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten


biological source

human

Quality Segment

assay

≥90% (SDS-PAGE)

form

lyophilized powder

specific activity

≥100 U/mg Protein, ≥50 U/mg Solid

mol wt

~63 kDa

suitability

suitable for molecular biology

UniProt accession no.

shipped in

wet ice

storage temp.

-10 to -25°C

Gene Information

human ... GPI(2821)

General description

PGI is a cytosolic enzyme that belongs to the isomerase class and catalyzes the reversible interconversion of glucose-6-phosphate and fructose-6-phosphate. It plays a central role in both glycolysis and gluconeogenesis. In addition to its intracellular metabolic function, PGI is also known to function extracellularly as a cytokine-like protein, including roles as autocrine motility factor (AMF) and neuroleukin, highlighting its multifunctional (“moonlighting”) properties.

Application

Phosphoglucose isomerase has been used in:
  • Glycolysis and metabolic pathway studies
  • Enzyme kinetics and activity assays
  • Cancer metabolism and cell migration studies
  • Functional studies of metabolic enzymes

Biochem/physiol Actions

PGI catalyzes the isomerization of glucose-6-phosphate to fructose-6-phosphate, a critical step in glycolysis. In addition to its metabolic role, it has extracellular signaling functions associated with tumor cell motility and growth regulation.

Physical form

Centrifuge the vial at 4ºC before opening to recover the entire contents. It is recommended that sterile water be added to the vial to prepare a stock solution.
Lyophilized from 10mM KPB (pH 8.0), 0.150M NaCl

Preparation Note

Centrifuge the vial at 4ºC before opening to recover the entire contents. It is recommended that sterile water be added to the vial to prepare a stock solution.
Unit definition: One enzyme activity unit is defined as the amount of enzyme required to convert 1.0 μmol of D-fructose-6-phosphate to D-glucose-6-phosphate per minute at pH 7.4 and a temperature of 25 °C.

Ähnliche Artikel vergleichen

Vollständigen Vergleich anzeigen

Unterschiede anzeigen

1 of 1

Dieser Artikel
G6430P9544PGIRO
description

Lyophilized powder, ≥ 100U/mg protein

description

Liquid, ≥ 14U/mg protein

description

Type XI, lyophilized powder, ≥200 units/mg protein

description

from yeast

biological source

human

biological source

human

biological source

rabbit muscle

biological source

yeast

assay

≥90% (SDS-PAGE)

assay

-

assay

-

assay

-

suitability

suitable for molecular biology

suitability

suitable for molecular biology

suitability

-

suitability

-

form

lyophilized powder

form

liquid

form

lyophilized powder

form

suspension

mol wt

~63 kDa

mol wt

~60 kDa

mol wt

-

mol wt

-

shipped in

wet ice

shipped in

wet ice

shipped in

-

shipped in

-


Piktogramme

Health hazard

Signalwort

Danger

Gefahrencodes

Sicherheitshinweise

Hazard Classifications

Resp. Sens. 1

Lagerklasse

11 - Combustible Solids



Hier finden Sie alle aktuellen Versionen:

Analysenzertifikate (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumente section.

Wenn Sie Hilfe benötigen, wenden Sie sich bitte an Kundensupport

Besitzen Sie dieses Produkt bereits?

In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.

Die Dokumentenbibliothek aufrufen