Direkt zum Inhalt
Merck

Fortfahren mit

S8840

Proteasehemmer-Cocktail

For general use, a broad-spectrum inhibitor cocktail

Synonym(e):

Protease Inhibitor Cocktail, Protein detection, Protein phosphorylation, Protein sample preparation

Anmelden zur Ansicht der Organisations- und Vertragspreise.

Größe auswählen

Ansicht ändern
Größe/SKUVerfügbarkeitPreis

Über diesen Artikel

UNSPSC Code:
12352200
MDL number:
Form:
tablet
Solubility:
soluble (clear to slightly hazy)
Storage temp.:
2-8°C
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten


biological source

bovine

Quality Segment

form

tablet

color

white

input

general use sample(s)

solubility

soluble (clear to slightly hazy)

storage temp.

2-8°C

General description

Unlock the power of protein phosphorylation with SIGMAFAST Phosphatase Inhibitor Tablets (S8840). Designed for researchers and scientists, this broad-spectrum inhibitor cocktail is essential for preserving the integrity of phosphorylated proteins in your cell extracts. Phosphorylation is a critical post-translational modification that regulates enzyme activity and triggers vital cellular signaling pathways.

Key Components:

  • Bromolevamisole oxalate: A potent alkaline phosphatase inhibitor.
  • Cantharidin: Selectively inhibits protein phosphatase 2 (PP2A).
  • Sodium fluoride: An irreversible inhibitor of serine, threonine, and acid phosphatases.
  • Sodium orthovanadate: Targets tyrosine and alkaline phosphatases.
How to Use:

To prepare your phosphatase inhibitor solution, simply dissolve one S8840 tablet in 20 mL of water or cell lysis buffer to generate a 1X solution. For more concentrated applications, reconstitute the tablet in a smaller volume (2 mL) to create a 10X stock solution, which can then be diluted as needed. Please note, concentrations greater than 1X may appear hazy, but this will not affect the performance of the inhibitors. Endogenous proteases are also present during cell lysis which could lead to proteolytic degradation of proteins. Supplementing the lysis buffer with protease inhibitors prevents any proteins of interest from being degraded in this fashion. Sigma Cat#S8830 is a protease inhibitor cocktail that can be used for this purpose.

Application

Ideal for research in cancer biology, cellular signaling, and enzyme activity studies.

Features and Benefits

  • Broad-spectrum inhibition: Protects against a wide range of phosphatases.
  • Easy preparation: Quick and straightforward dilution process.
  • High stability: Reliable performance for extended periods.
  • Effectively Preserves the integrity of phosphorylated proteins
  • Convenient fast dissolving tablet, saves time by eliminating powder weigh-upTablet is stable for 4 years

Preparation Note

  • Unopened tablets are stable for up to 4 years at 2-8 °C.
  • Once reconstituted, the inhibitor solution remains stable for approximately 2 weeks at 2-8°C.
  • Avoid freezing the reconstituted solutions to prevent precipitation.

Ähnliche Artikel vergleichen

Vollständigen Vergleich anzeigen

Unterschiede anzeigen

1 of 1

Dieser Artikel
S8850S8820S8830
description

For general use, a broad-spectrum inhibitor cocktail

description

-

description

For General Use

description

for use in purification of Histidine-tagged proteins

form

tablet

form

tablet

form

tablet

form

tablet

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

biological source

bovine

biological source

bovine

biological source

bovine lung, bovine milk

biological source

bovine lung

solubility

soluble (clear to slightly hazy)

solubility

soluble (clear to slightly hazy)

solubility

water: 1 tablet/100mL, clear to hazy, colorless

solubility

-

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

color

white

color

white

color

-

color

-


Piktogramme

Exclamation mark

Signalwort

Warning

Gefahrencodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

supp_hazards

Lagerklasse

11 - Combustible Solids

Was ist los?

WGK 3



Hier finden Sie alle aktuellen Versionen:

Analysenzertifikate (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumente section.

Wenn Sie Hilfe benötigen, wenden Sie sich bitte an Kundensupport

Besitzen Sie dieses Produkt bereits?

In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.

Die Dokumentenbibliothek aufrufen