Über diesen Artikel
Empirische Formel (Hill-System):
C18H14ClFN4O
CAS-Nummer:
Molekulargewicht:
356.78
UNSPSC Code:
12352200
PubChem Substance ID:
MDL number:
eCl@ss:
32150313
Assay:
≥98% (HPLC)
Form:
powder
assay
≥98% (HPLC)
form
powder
drug control
regulated under CDSA - not available from Sigma-Aldrich Canada
color
, white to yellow to light brown
solubility
DMSO: 20 mg/mL, clear
storage temp.
2-8°C
SMILES string
ClC1=CC=CC=C1C2=NC3=C(C)NN=C3NC4=C2C=C(F)C(OC)=C4
InChI
1S/C18H14ClFN4O/c1-9-16-18(24-23-9)21-14-8-15(25-2)13(20)7-11(14)17(22-16)10-5-3-4-6-12(10)19/h3-8H,1-2H3,(H2,21,23,24)
InChI key
UOVCGJXDGOGOCZ-UHFFFAOYSA-N
Gene Information
human ... AURKA(6790), FGFR1(2260), FGFR2(2263), FLT1(2321), FLT3(2322), FLT4(2324), KDR(3791), PDGFRB(5159), PLK4(10733)
Biochem/physiol Actions
R1530 is a multi-kinase inhibitor and an anti tumor agent.
R1530 is a multi-kinase inhibitor with activity against several kinases associated with tumor growth and angiogenesis, including VEGFR, FGFR, PDGFR, Chk2 and Cdk2. In tumor cells, R1530 inhibits tubulin polyerization and mitotic checkpoint function, resulting in polyploidy. The compound R1530 inhibits tumor growth and angiogenesis in animal models.
Signalwort
Warning
Gefahrencodes
Sicherheitshinweise
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Zielorgane
Respiratory system
Lagerklasse
11 - Combustible Solids
WGK
WGK 3
Flammpunkt (°F)
Not applicable
Flammpunkt (°C)
Not applicable
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?