Direkt zum Inhalt
Merck

Fortfahren mit

Z-2197

ZytoLight® SPEC PDGFRB Dual Color Break Apart Probe

Synonym(e):

FISH probe, dual color FISH probe

Anmelden zur Ansicht der Organisations- und Vertragspreise.

Größe auswählen

Ansicht ändern
Größe/SKUVerfügbarkeitPreis

Über diesen Artikel

eCl@ss:
32039090
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten


product line

ZytoVision

packaging

pkg of 0.05 mL, pkg of 5 tests

manufacturer/tradename

ZytoVision

storage condition

protect from light

color

green to orange

application(s)

research use

storage temp.

2-8°C

General description

The ZytoLight® SPEC PDGFRB Dual Color Break Apart Probe (PL155) is a direct-labeled FISH probe for the qualitative detection of translocations of the human PDGFRB gene at 5q32 in cytologic specimens, using fluorescence in situ hybridization (FISH). The probe is intended for use with the ZytoLight FISH-Cytology Implementation Kit.

ZytoLight probes enable the precise identification of genetic abnormalities, including translocations, deletions, amplifications, and chromosomal aneuploidies, in formalin-fixed, paraffin-embedded (FFPE) tissue sections or cytology specimens, without the need for additional antibody-based detection.



Components

The ZytoLight SPEC PDGFRB Dual Color Break Apart Probe comprises:
  • ZyGreen (excitation 503 nm/emission 528 nm) labeled polynucleotides (~10.0 ng/µl), targeting sequences mapping in 5q32-q33.1* (chr5:149,548,518-150,285,722) distal to the PDGFRB breakpoint region;
  • ZyOrange (excitation 547 nm/emission 572 nm) labeled polynucleotides (~4.5 ng/µl), targeting sequences mapping in 5q32* (chr5:149,058,515-149,471,369) proximal to the PDGFRB breakpoint region;
  • Formamide based hybridization buffer
*according to Human Genome Assembly GRCh37/hg19



Principle

The fluorescence in situ hybridization (FISH) method enables the identification and visualization of particular nucleic acid sequences within cell samples. DNA fragments labeled with fluorescent dyes, known as FISH probes, are co-denatured with their complementary target DNA strands in the sample and then allowed to hybridize. Following hybridization, non-specific and unbound probe fragments are removed through stringency washes. The DNA is then counterstained with DAPI, and the hybridized probes are detected using a fluorescence microscope equipped with filters matched to the specific fluorochromes used to label the FISH probes.

Application

For the qualitative detection of translocations involving the human PDGFRB gene at 5q32 by fluorescence in situ hybridization (FISH)

Analysis Note

Using suitable filter sets, the probe hybridization signals appear green away from the PDGFRB breakpoint region and orange near it.
  • Normal scenario: in normal cells during interphase or cells without a PDGFRB gene region translocation, two green/orange fusion signals are observed.
  • Aberrant scenario: one translocated PDGFRB gene region is shown by one separate green and orange signal each.
Overlapping signals may appear yellow.

Legal Information

ZytoLight is a registered trademark of ZytoVision GmbH in Germany


Piktogramme

Health hazard

Signalwort

Danger

Gefahrencodes

Hazard Classifications

Carc. 2 - Repr. 1B - STOT RE 2 Oral

Zielorgane

Blood

Lagerklasse

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Zulassungslistungen

Zulassungslistungen werden hauptsächlich für chemische Produkte erstellt. Für nicht-chemische Produkte können hier nur begrenzte Angaben gemacht werden. Kein Eintrag bedeutet, dass keine der Komponenten gelistet ist. Es liegt in der Verantwortung des Benutzers, die sichere und legale Verwendung des Produkts zu gewährleisten.

75-12-7

CAS No.

75-12-7

CAS No.


Hier finden Sie alle aktuellen Versionen:

Analysenzertifikate (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumente section.

Wenn Sie Hilfe benötigen, wenden Sie sich bitte an Kundensupport

Besitzen Sie dieses Produkt bereits?

In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.

Die Dokumentenbibliothek aufrufen