Fortfahren mit
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Größe auswählen
Ansicht ändern
| Größe/SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
eCl@ss:
32160703
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltenproduct line
ZytoVision
manufacturer/tradename
ZytoVision
storage condition
protect from light
color
, Green/Orange/Blue
application(s)
research use
storage temp.
2-8°C
General description
The ZytoLight® SPEC PDGFRA/FIP1L1 TriCheck Probe (PL167) is a direct-labeled FISH probe designed for the qualitative detection of rearrangements involving the human PDGFRA gene, with and without participation from the FIP1L1 gene, in cytological specimens. The probe utilizes direct-labeled FISH technology, eliminating the need for fluorophore-coupled antibodies and is intended for use in a certified laboratory environment.
Components:
The ZytoLight SPEC PDGFRA/FIP1L1 TriCheck Probe is composed of:
The ZytoLight® SPEC PDGFRA/FIP1L1 TriCheck Probe uses direct-labeled FISH probes that hybridize to specific target DNA sequences in the 4q12 chromosomal region of cell preparations. Fluorescently-labeled DNA fragments and their complementary target DNA strands are co-denatured and allowed to anneal during hybridization. Unbound probe fragments are removed by stringency washing steps. After counterstaining with DAPI, hybridized probes are visualized using a fluorescence microscope equipped with excitation and emission filters specific for ZyGreen (503/528 nm), ZyOrange (547/572 nm), and ZyBlue (418/467 nm) fluorochromes.
Components:
The ZytoLight SPEC PDGFRA/FIP1L1 TriCheck Probe is composed of:
- ZyGreen labeled polynucleotides (~10.0 ng/µl).
- ZyOrange labeled polynucleotides (~4.5 ng/µl).
- ZyBlue labeled polynucleotides (~37.0 ng/µl).
- Formamide-based hybridization buffer.
The ZytoLight® SPEC PDGFRA/FIP1L1 TriCheck Probe uses direct-labeled FISH probes that hybridize to specific target DNA sequences in the 4q12 chromosomal region of cell preparations. Fluorescently-labeled DNA fragments and their complementary target DNA strands are co-denatured and allowed to anneal during hybridization. Unbound probe fragments are removed by stringency washing steps. After counterstaining with DAPI, hybridized probes are visualized using a fluorescence microscope equipped with excitation and emission filters specific for ZyGreen (503/528 nm), ZyOrange (547/572 nm), and ZyBlue (418/467 nm) fluorochromes.
Application
For the qualitative detection of human PDGFRA rearrangements with and without participation of the human FIP1L1 gene by fluorescence in situ hybridization (FISH)
Analysis Note
The hybridization signals from the probe are colored blue (distal to the PDGFRA gene region), orange (proximal to the PDGFRA gene region), and green (proximal to the FIP1L1 gene region). Expected patterns include:Normal situation: Two green/orange fusion signals appear for specimens without rearrangements; when using a triple bandpass filter set, merged signals may be observed.Aberrant situation: Fusion events can be identified by variations in signal patterns, such as the presence of separate green and blue signals or a loss of signals indicating potential rearrangements.
Legal Information
ZytoLight is a registered trademark of ZytoVision GmbH in Germany
Signalwort
Danger
Gefahrencodes
Sicherheitshinweise
Hazard Classifications
Carc. 2 - Repr. 1B - STOT RE 2 Oral
Zielorgane
Blood
Lagerklasse
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
Zulassungslistungen
Zulassungslistungen werden hauptsächlich für chemische Produkte erstellt. Für nicht-chemische Produkte können hier nur begrenzte Angaben gemacht werden. Kein Eintrag bedeutet, dass keine der Komponenten gelistet ist. Es liegt in der Verantwortung des Benutzers, die sichere und legale Verwendung des Produkts zu gewährleisten.
75-12-7
CAS No.
75-12-7
CAS No.
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?
In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.
