Direkt zum Inhalt
Merck

Fortfahren mit

1A00130

USP

QUETIAPINE TETRAETHYLENE GLYCOL FUMARATE SALT

Pharmaceutical Analytical Impurity (PAI)

Synonym(e):

Quetiapine Tetraethylene Glycol Fumarate Salt, (2-(2-(2-(2-(4-(dibenzo[b,f][1,4]thiazepin-11-yl)piperazin-1-yl)ethoxy)ethoxy)ethoxy)ethan-1-ol-fumarate)

Anmelden zur Ansicht der Organisations- und Vertragspreise.

Größe auswählen

Ansicht ändern
Größe/SKUVerfügbarkeitPreis

Über diesen Artikel

Empirische Formel (Hill-System):
C25H33N3O4S·C4H4O4
CAS-Nummer:
Molekulargewicht:
587.68
MDL number:
NACRES:
NA.24
UNSPSC Code:
41116107
eCl@ss:
25140904
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhalten


grade

pharmaceutical analytical impurity (PAI)

agency

USP

API family

quetiapine

manufacturer/tradename

USP

application(s)

pharmaceutical
pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

2-8°C

General description

Find answers to frequently asked questions on USP products in USP FAQs.
QUETIAPINE TETRAETHYLENE GLYCOL FUMARATE SALT is a USP Pharmaceutical Analytical Impurity (PAI).
USP PAI are a product line of impurities suitable for research and analytical purposes, which help to ensure the quality and safety of medicines.
Associated Drug Substance: Quetiapine fumarate
Therapeutic Area: Psychiatrics

Application

QUETIAPINE TETRAETHYLENE GLYCOL FUMARATE SALT (USP PAI) is intended for use in analytical testing to detect, identify, and measure pharmaceutical impurities.

Features and Benefits

USP PAI advance your early analytical R&D and process development. PAI can be used in the following applications:
1. Conduct analytical tests during early formulation feasibility studies.
2. Determine degradation impurities produced during stress studies.
3. Develop, validate, and transfer analytical methods.
4. Perform spiking studies during process R&D to demonstrate depletion upon recrystallization.
5. Record retention times and/or spectra
6. Determine relative response factors.
7. Identify unknown impurities that formed during ICH stability conditions.
8. Identify impurities that are present in the Reference Listed Drug
9. Test for and profile impurities not listed in drug substance and drug product monographs.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

Sales restrictions may apply.
USP issued SDS can be found here

Ähnliche Artikel vergleichen

Vollständigen Vergleich anzeigen

Unterschiede anzeigen

1 of 1

Dieser Artikel
1592704Y0001657PHR1856
description

Pharmaceutical Analytical Impurity (PAI)

description

United States Pharmacopeia (USP) Reference Standard, monograph mol wt. 883.09 ((C21H25N3O2S)2 · C4H4O4)

description

European Pharmacopoeia (EP) Reference Standard

description

Pharmaceutical Secondary Standard; Certified Reference Material

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

EDQM

manufacturer/tradename

-

grade

pharmaceutical analytical impurity (PAI)

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical secondary standard, certified reference material

application(s)

pharmaceutical
pharmaceutical
pharmaceutical small molecule

application(s)

pharmaceutical
pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)
pharmaceutical

application(s)

pharmaceutical

format

neat

format

neat

format

neat

format

-

agency

USP

agency

USP

agency

-

agency

traceable to USP 1592704, traceable to Ph. Eur. Y0006157

API family

quetiapine

API family

quetiapine

API family

quetiapine

API family

-


Lagerklasse

11 - Combustible Solids



Hier finden Sie alle aktuellen Versionen:

Analysenzertifikate (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumente section.

Wenn Sie Hilfe benötigen, wenden Sie sich bitte an Kundensupport

Besitzen Sie dieses Produkt bereits?

In der Dokumentenbibliothek finden Sie die Dokumentation zu den Produkten, die Sie kürzlich erworben haben.

Die Dokumentenbibliothek aufrufen