Merck
A new approach to the analysis of nicarbazin and ionophores in eggs by HPLC/MS/MS.
Journal of AOAC International (2011-05-14)
Jasna Dmitrovic, David A Durden
ZUSAMMENFASSUNG
An HPLC/MS/MS method has been developed and validated for the quantification and confirmation of nicarbazin and ionophores (lasalocid, monensin, salinomycin, and narasin) in eggs. Nicarbazin is determined in the negative electrospray mode with a basic mobile phase that supports creation of negative ions. Consequently, our ability to maintain instrument sensitivity over time has significantly improved. The analysis of the ionophores is done in the positive electrospray mode using ammonium buffer for HPLC separation. Monitoring ammonium adduct parent ions resulted in enhanced sensitivity and better reproducibility of the ionophore analysis. The validation of this improved HPLC/MS/MS method for the detection of nicarbazin and the ionophores demonstrated excellent precision of below 10% RSD and lower LOD values (microg/kg) for nicarbazin (0.018), lasalocid (0.015), monensin (0.015), salinomycin (0.033), and narasin (0.039).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monensin Natriumsalz, 90-95% (TLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Nicarbazin, VETRANAL®, analytical standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Nicarbazin
Preise und Verfügbarkeit sind derzeit nicht verfügbar.