Merck
  • CBX8, a component of the Polycomb PRC1 complex, modulates DOT1L-mediated gene expression through AF9/MLLT3.
CBX8, a component of the Polycomb PRC1 complex, modulates DOT1L-mediated gene expression through AF9/MLLT3.
FEBS letters (2013-07-31)
Bhavna Malik, Charles S Hemenway
ZUSAMMENFASSUNG
AF9 is known to interact with multiple proteins including activators and repressors of transcription. Our data indicate that other AF9 binding proteins compete with the histone methyltransferase DOT1L for AF9 binding thus diminishing its ability to methylate lysine 79 of histone 3. Specifically, we show that AF9 is part of a protein multimer containing members of Polycomb group (PcG) PRC1 complex, CBX8, RING1B, and BMI1. Interaction with CBX8 precludes AF9-DOT1L binding. Knockdown of CBX8 with short-hairpin RNA (shRNA) leads to decreased expression of the AF9 target gene ENaCα. In contrast, CBX8 overexpression results in increased ENaCα mRNA levels and this effect can be partially overcome by co-overexpression of AF9.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler ANTI-FLAG® M2-Antikörper in Maus hergestellte Antikörper, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-HA-Antikörper, monoklonaler Antikörper der Maus in Maus hergestellte Antikörper, clone HA-7, purified from hybridoma cell culture
Sigma-Aldrich
Chromatin-Immunpräzipitations-Kit EZ-Magna ChIP® A/G, Single day chromatin immunoprecipitation (ChIP) kit containing all necessary reagents to perform 22 individual chromatin immunoprecipitation (ChIP) reactions using magnetic A/G beads. Control primers included.
Sigma-Aldrich
Anti-Myc-Tag-Antikörper, Upstate®, from rabbit