Merck
  • Examination of the Human Cytochrome P4503A4 Induction Potential of PF-06282999, an Irreversible Myeloperoxidase Inactivator: Integration of Preclinical, In Silico, and Biomarker Methodologies in the Prediction of the Clinical Outcome.
Examination of the Human Cytochrome P4503A4 Induction Potential of PF-06282999, an Irreversible Myeloperoxidase Inactivator: Integration of Preclinical, In Silico, and Biomarker Methodologies in the Prediction of the Clinical Outcome.
Drug metabolism and disposition: the biological fate of chemicals (2017-03-04)
Jennifer Q Dong, James R Gosset, Odette A Fahmi, Zhiwu Lin, Jeffrey R Chabot, Steven G Terra, Vu Le, Kristin Chidsey, Parya Nouri, Albert Kim, Leonard Buckbinder, Amit S Kalgutkar
ZUSAMMENFASSUNG
The propensity for CYP3A4 induction by 2-(6-(5-chloro-2-methoxyphenyl)-4-oxo-2-thioxo-3,4-dihydropyrimidin-1(2
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
PF-06282999, ≥98% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.