Merck
  • Tetracyclines, sulfonamides and quinolones and their corresponding resistance genes in the Three Gorges Reservoir, China.
Tetracyclines, sulfonamides and quinolones and their corresponding resistance genes in the Three Gorges Reservoir, China.
The Science of the total environment (2018-05-08)
Muting Yan, Chen Xu, Yumei Huang, Huayue Nie, Jun Wang
ZUSAMMENFASSUNG
The Three Gorges Project significantly impacted water quality and ecological balance in this area. The special engineered aquatic environment could be an important reservoir for antibiotic resistance genes (ARGs). Fifteen ARGs corresponding to three groups of antibiotics (tetracyclines, sulfonamides and quinolones) were determined in surface water, soil and sediment in this study. Total concentrations of antibiotics ranged from 21.55 to 536.86ng/L, 3.69 to 438.76ng/g, 15.78 to 213.84ng/g in water, soil and sediment, respectively. Polymerase chain reaction (PCR) of ARGs revealed the presence of two sulfonamide resistance genes (sul1, sul2), five tetracycline resistance genes (tetA, tetB, tetM, tetQ, tetG) and class 1 integron gene (intI1) in all samples. And the relative abundance of sulfonamide resistance genes was generally higher than tetracycline resistance genes in three matrices. Significant correlations (p<0.05) were found between the concentrations of intI1 and ARGs (tetA, tetB, tetM, tetQ, tetG, sul1, sul2), indicating intI1 may facilitate the proliferation and propagation of these genes. Redundancy analysis (RDA) showed distribution of ARGs was related to the certain antibiotics residues, which may exert selective pressure on bacteria and thus enrich the abundance of ARGs. The results of this study could provide useful information for both better understanding and management of the contamination caused by ARGs and related antibiotics in engineered aquatic environments.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N-Octanoyl-N-Methylglucamin, ≥97% (GC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.