Merck
  • Synthesis of New C- and N-β-d-Glucopyranosyl Derivatives of Imidazole, 1,2,3-Triazole and Tetrazole, and Their Evaluation as Inhibitors of Glycogen Phosphorylase.
Synthesis of New C- and N-β-d-Glucopyranosyl Derivatives of Imidazole, 1,2,3-Triazole and Tetrazole, and Their Evaluation as Inhibitors of Glycogen Phosphorylase.
Molecules (Basel, Switzerland) (2018-03-16)
Sándor Kun, Éva Bokor, Ádám Sipos, Tibor Docsa, László Somsák
ZUSAMMENFASSUNG
The aim of the present study was to broaden the structure-activity relationships of C- and N-β-d-glucopyranosyl azole type inhibitors of glycogen phosphorylase. 1-Aryl-4-β-d-gluco-pyranosyl-1,2,3-triazoles were prepared by copper catalyzed azide-alkyne cycloadditions between O-perbenzylated or O-peracetylated β-d-glucopyranosyl ethynes and aryl azides. 1-β-d-Gluco-pyranosyl-4-phenyl imidazole was obtained in a glycosylation of 4(5)-phenylimidazole with O-peracetylated α-d-glucopyranosyl bromide. C-β-d-Glucopyranosyl-N-substituted-tetrazoles were synthesized by alkylation/arylation of O-perbenzoylated 5-β-d-glucopyranosyl-tetrazole or from a 2,6-anhydroheptose tosylhydrazone and arenediazonium salts. 5-Substituted tetrazoles were glycosylated by O-peracetylated α-d-glucopyranosyl bromide to give N-β-d-glucopyranosyl-C-substituted-tetrazoles. Standard deprotections gave test compounds which were assayed against rabbit muscle glycogen phosphorylase b. Most of the compounds proved inactive, the best inhibitor was 2-β-d-glucopyranosyl-5-phenyltetrazole (IC50 600 μM). These studies extended the structure-activity relationships of β-d-glucopyranosyl azole type inhibitors and revealed the extreme sensitivity of such type of inhibitors towards the structure of the azole moiety.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pyridin, anhydrous, 99.8%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.