Merck
  • Meta-analysis identifies five novel loci associated with endometriosis highlighting key genes involved in hormone metabolism
Meta-analysis identifies five novel loci associated with endometriosis highlighting key genes involved in hormone metabolism
Nature Communications (2017)
Sapkota Y, et al.