Merck
  • Neuronal nitric oxide synthase immunohistochemistry and 4, 5-diaminofluorescein diacetate: tools for nitric oxide research.
Neuronal nitric oxide synthase immunohistochemistry and 4, 5-diaminofluorescein diacetate: tools for nitric oxide research.
Journal of Neuroscience Methods (2001)
Kasim N, et al.