Merck
Manipulation of facet orientation in hybrid perovskite polycrystalline films by cation cascade.
Nature communications (2018-07-20)
Guanhaojie Zheng, Cheng Zhu, Jingyuan Ma, Xiaonan Zhang, Gang Tang, Runguang Li, Yihua Chen, Liang Li, Jinsong Hu, Jiawang Hong, Qi Chen, Xingyu Gao, Huanping Zhou
ZUSAMMENFASSUNG
Crystal orientations in multiple orders correlate to the properties of polycrystalline materials, and it is critical to manipulate these microstructural arrangements to enhance device performance. Herein, we report a controllable approach to manipulate the facet orientation within the ABX3 hybrid perovskites polycrystalline films by cation cascade doping at A-site. Two-dimensional synchrotron radiation grazing incidence wide-angle X-ray scattering is employed to probe the crystal orientations in multiple orders in mixed perovskites thin films, revealing a general pattern to guide crystal planes stacking upon extrinsic doping during crystallization. Different from previous studies, this method enables to adjust the crystal stacking mode of certain crystallographic planes in polycrystalline perovskites. Moreover, the preferred facet orientation is found to facilitate photocarrier transport across the absorber and pertaining interface in the resultant PV device, which provides an exemplary paradigm for further explorations that relate to the microstructures of hybrid perovskite materials and relevant optoelectronics.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Chlorbenzol, suitable for HPLC, 99.9%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
4-tert-Butylpyridin, 98%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.