Merck
  • Inhibitory Effect of Persimmon Tannin on Pancreatic Lipase and the Underlying Mechanism in Vitro.
Inhibitory Effect of Persimmon Tannin on Pancreatic Lipase and the Underlying Mechanism in Vitro.
Journal of agricultural and food chemistry (2018-05-29)
Wei Zhu, Yangyang Jia, Jinming Peng, Chun-Mei Li
ZUSAMMENFASSUNG
Pancreatic lipase (PL) is a critical enzyme associated with hyperlipidemia and obesity. A previous study of ours suggested that persimmon tannin (PT) was the main component accounting for the antihyperlipidemic effects of persimmon fruits, but the underlying mechanisms were unclear. In this present study, the inhibitory effect of PT on PL was studied and the possible mechanisms were evaluated by fluorescence spectroscopy, circular dichroism (CD) spectra, isothermal titration calorimetry (ITC), and molecular docking. PT had a high affinity to PL and inhibited the activity of PL with the half maximal inhibitory concertation (IC50) value of 0.44 mg/mL in a noncompetitive way. Furthermore, molecular docking revealed that the hydrogen bonding and π-π stacking was mainly responsible for the interaction. The strong inhibition of PT on PL in the gastrointestinal tract might be one mechanism for its lipid-lowering effect.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Lipase aus Schweinepankreas, Type II, ≥125 units/mg protein (using olive oil (30 min incubation)), 30-90 units/mg protein (using triacetin)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.