Merck
Signaling by TGF-betas in tubule cultures of adult rat testis.
American journal of translational research (2017-04-08)
Kai-Hui Chan, Sebastian P Galuska, Pradeep Kumar Kudipudi, Mohammad Assad Riaz, Kate L Loveland, Lutz Konrad
ZUSAMMENFASSUNG
Although signal transduction of transforming growth factor-betas (TGF-βs) is well characterized in individual cell types, data about TGF-β signaling in a cellular context is still scarce. In this study, we used ex vivo tubule cultures from adult rat testis to investigate TGF-β signaling. We show for the first time in testicular tubules, that TGF-βs also signal via the BMP type I receptors, with ALK2 used by TGF-β1 and ALK3 and ALK6 by TGF-β2. This signal transduction is mediated via Smad3 as well as via Smad1. In contrast, BMPs (BMP2 and BMP7) do not signal via the high-affinity type I and type II TGFβ receptors, TBR1 or TBR2. Furthermore, treatment of tubule cultures with either TGF-β1 or TGF-β2 had profound significant stimulatory effects on secretion of plasminogen activator-1 (PAI-1) through utilization of TGF-β and BMP receptors. Specific inhibitors for either TBR1 or BMP receptors yielded nearly complete inhibition of TGF-β signaling. The TBR1-TBR2 signalosome was detected with Duolink upon stimulation with either TGF-β1 or TGF-β2, predominantly in spermatogenic cells of the adult rat testis, particularly in elongated spermatids. In summary, this examination of intact rat testicular tubules demonstrated for the first time that TGF-βs signal mainly through TBR1 and TBR2 but also use BMP receptors, including for secretion of PAI-1. Whereas ALK2 participates in the TGF-β1-induced TBR1-TBR2 signalosome, ALK3 and ALK6 are involved in signaling of TGF-β2. Detection of the TBR1-TBR2 signalosome in late spermiogenic cells indicates a post-meiotic activity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Duolink® In Situ Detektionsreagenzien Rot
Sigma-Aldrich
Duolink® In Situ PLA® Sonde Anti-Kaninchen PLUS, Affinity purified Donkey anti-Rabbit IgG (H+L)
Sigma-Aldrich
Duolink® In Situ Rot Starterkit Maus/Kaninchen
Sigma-Aldrich
Duolink® In Situ PLA® Sonde Anti-Maus MINUS, Affinity purified Donkey anti-Mouse IgG (H+L)
Sigma-Aldrich
Duolink® In Situ PLA® Sonde Anti-Maus PLUS
Sigma-Aldrich
Duolink® In Situ PLA® Sonde Anti-Kaninchen MINUS
Sigma-Aldrich
Duolink® In-situ-Detektionsreagenzien Orange
Sigma-Aldrich
Duolink® In-situ-Detektionsreagenzien Tiefrot
Sigma-Aldrich
Duolink® In-situ-Detektionsreagenzien Grün
Sigma-Aldrich
Duolink® In Situ Waschpuffer, Fluoreszenz
Sigma-Aldrich
Duolink® In Situ Orange Starterkit Maus/Kaninchen
Sigma-Aldrich
Duolink® In Situ Eindeckmittel mit DAPI
Sigma-Aldrich
Duolink® In Situ Probemaker PLUS
Sigma-Aldrich
Duolink® In Situ Probemaker MINUS
Sigma-Aldrich
Duolink® In Situ Detection Reagents Brightfield
Sigma-Aldrich
Duolink® flowPLA Detection Kit - FarRed, Duolink® PLA kit for Flow Cytometry with FarRed Detection
Sigma-Aldrich
Duolink® PLA Control Kit - PPI, Control Kit to reliably detect Protein-Protein Interactions (PPI) with Duolink® PLA
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat MINUS, Affinity purified Donkey anti-Goat IgG (H+L)
Sigma-Aldrich
Duolink® In Situ Rot Starterkit Ziege/Kaninchen
Sigma-Aldrich
Duolink® In Situ Wash Buffer, Brightfield
Sigma-Aldrich
Duolink® In Situ PLA® Sonde Anti-Ziege PLUS
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Red, DUOLINK® : PLA kit for Flow Cytometry with Red Detection
Sigma-Aldrich
Duolink®-flowPLA-Detektionskit – Grün, Duolink® PLA kit for Flow Cytometry with Green Detection
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Goat
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Goat/Rabbit
Sigma-Aldrich
Duolink® In Situ Microplate Nuclear Stain, Anti-Fade
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Orange, Duolink® PLA kit for Flow Cytometry with Orange Detection
Sigma-Aldrich
Duolink® In Situ Microplate Heat Transfer Block
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Mouse/Goat