Merck
  • Fracture risk in type 2 diabetic patients: A clinical prediction tool based on a large population-based cohort.
Fracture risk in type 2 diabetic patients: A clinical prediction tool based on a large population-based cohort.
PloS one (2018-09-08)
Daniel Martínez-Laguna, Cristian Tebé, Xavier Nogués, M Kassim Javaid, Cyrus Cooper, Victor Moreno, Adolfo Diez-Perez, Gary S Collins, Daniel Prieto-Alhambra
ZUSAMMENFASSUNG
An increased fracture risk has been described as a complication of Type 2 diabetes mellitus (T2DM). Clinical prediction models for general population have a limited predictive accuracy for fractures in T2DM patients. The aim was to develop and validate a clinical prediction tool for the estimation of 5-year hip and major fracture risk in T2DM patients. A cohort of newly diagnosed T2DM patients (n = 51,143, aged 50-85, 57% men) was extracted from the Information System for the Development of Research in Primary Care (SIDIAP) database, containing computerized primary care records for >80% of the population of Catalonia, Spain (>6 million people). Patients were followed up from T2DM diagnosis until the earliest of death, transfer out, fracture, or end of study. Cox proportional hazards regression was used to model the 5-year risk of hip and major fracture. Calibration and discrimination were assessed. Hip and major fracture incidence rates were 1.84 [95%CI 1.64 to 2.05] and 7.12 [95%CI 6.72 to 7.53] per 1,000 person-years, respectively. Both hip and major fracture prediction models included age, sex, previous major fracture, statins use, and calcium/vitamin D supplements; previous ischemic heart disease was also included for hip fracture and stroke for major fracture. Discrimination (0.81 for hip and 0.72 for major fracture) and calibration plots support excellent internal validity. The proposed prediction models have good discrimination and calibration for the estimation of both hip and major fracture risk in incident T2DM patients. These tools incorporate key T2DM macrovascular complications generally available in primary care electronic medical records, as well as more generic fracture risk predictors. Future work will focus on validation of these models in external cohorts.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-(Diisopropylamino)ethylmethacrylat, 97%, contains ~100 ppm monomethyl ether hydroquinone as inhibitor
Preise und Verfügbarkeit sind derzeit nicht verfügbar.