- MS-based conformation analysis of recombinant proteins in design, optimization and development of biopharmaceuticals.
MS-based conformation analysis of recombinant proteins in design, optimization and development of biopharmaceuticals.
Methods (San Diego, Calif.) (2018-04-22)
Devrishi Goswami, Jun Zhang, Pavel V Bondarenko, Zhongqi Zhang
PMID29678586
ZUSAMMENFASSUNG
Mass spectrometry (MS)-based methods for analyzing protein higher order structures have gained increasing application in the field of biopharmaceutical development. The predominant methods used in this area include native MS, hydrogen deuterium exchange-MS, covalent labeling, cross-linking and limited proteolysis. These MS-based methods will be briefly described in this article, followed by a discussion on how these methods contribute at different stages of discovery and development of protein therapeutics.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Preise und Verfügbarkeit sind derzeit nicht verfügbar.