Merck
  • Development and validation of HPLC methods for analysis of chlorantraniliprole insecticide in technical and commercial formulations.
Development and validation of HPLC methods for analysis of chlorantraniliprole insecticide in technical and commercial formulations.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes (2018-03-07)
Mohamed E I Badawy
ZUSAMMENFASSUNG
Effective, selective, precise and accurate liquid chromatographic analytical methods for the analysis of a novel chlorantraniliprole insecticide in technical and formulation (coragen, 20% SC) have been optimized and validated. Eight methods were designed based on different mobile phases, temperature and two HPLC columns. The mobile phase consists of two mixtures (acetonitrile:water, 70:30 and methanol:water, 70:30) with 25 or 40ºC. HPLC analysis of chlorantraniliprole was carried out at a wavelength of 260 nm, with a flow rate of 0.8 mL/min. The calibration curves showed a good linear relationship (R2 ˃ 0.99) in the injected quantities ranged from 0.0125 to 1.00 μg. Limit of detection (LOD) was found to be 3.94 to 14.56 ng and from 5.95 to 12.93 ng using the analytical methods I to IV by MicroPack CN-10 and V-VIII by ZORBAX Eclips Plus C18 columns, respectively, based on SDslope values. ZORBAX Eclips Plus C18 column with method VI was the best one (R2 = 1.00 and RSD = 0.30), short retention time (4.936 min), high theoretical plates per column (65457.15) compared to others and LOD = 6.49 ng. The accuracy of the best method was demonstrated by recovery rates of 83.04% to 98.50% for grape samples supplemented with 5, 10 and 50 mg chlorantraniliprole/kg.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Chlorantraniliprol, PESTANAL®, analytical standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.