Merck
  • An engineered thermal-shift screen reveals specific lipid preferences of eukaryotic and prokaryotic membrane proteins.
An engineered thermal-shift screen reveals specific lipid preferences of eukaryotic and prokaryotic membrane proteins.
Nature communications (2018-10-14)
Emmanuel Nji, Yurie Chatzikyriakidou, Michael Landreh, David Drew
ZUSAMMENFASSUNG
Membrane bilayers are made up of a myriad of different lipids that regulate the functional activity, stability, and oligomerization of many membrane proteins. Despite their importance, screening the structural and functional impact of lipid-protein interactions to identify specific lipid requirements remains a major challenge. Here, we use the FSEC-TS assay to show cardiolipin-dependent stabilization of the dimeric sodium/proton antiporter NhaA, demonstrating its ability to detect specific protein-lipid interactions. Based on the principle of FSEC-TS, we then engineer a simple thermal-shift assay (GFP-TS), which facilitates the high-throughput screening of lipid- and ligand- interactions with membrane proteins. By comparing the thermostability of medically relevant eukaryotic membrane proteins and a selection of bacterial counterparts, we reveal that eukaryotic proteins appear to have evolved to be more dependent to the presence of specific lipids.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Guanosin-5′-Monophosphat Dinatriumsalz Hydrat, from yeast, ≥99%
Sigma-Aldrich
Cytidine 5′-monophosphate disodium salt, Sigma Grade, ≥99%, crystalline
Sigma-Aldrich
Uridin 5′-monophosphat Dinatriumsalz, ≥99%
Sigma-Aldrich
N-Acetylneuraminsäure, ≥95% anhydrous basis, synthetic
Sigma-Aldrich
Cytidin-5′-Monophospho-N-Acetylneuraminsäure Natriumsalz, ≥85% (HPLC)
Sigma-Aldrich
Cholesterylhemisuccinat Tris-salz, anionic detergent
Sigma-Aldrich
Uridine 5′-monophosphate, ≥98%
Sigma-Aldrich
Uridin-5′-diphosphoglucose Dinatriumsalz, ≥98.0% (HPLC)
Sigma-Aldrich
L-α-Phosphatidylethanolamin, Dioleoyl, ≥99% (GC), ≥98% (TLC), lyophilized powder
Sigma-Aldrich
Cytidin 5′-Diphosphocholin Natriumsalz Dihydrat, ~98%, from yeast, solid
Sigma-Aldrich
Brain Extract from bovine brain, Type I, Folch Fraction I