Merck
  • Mouse tissues that undergo neoplastic progression after K-Ras activation are distinguished by nuclear translocation of phospho-Erk1/2 and robust tumor suppressor responses.
Mouse tissues that undergo neoplastic progression after K-Ras activation are distinguished by nuclear translocation of phospho-Erk1/2 and robust tumor suppressor responses.
Molecular cancer research : MCR (2012-04-26)
Neha Parikh, Ryan L Shuck, Thuy-Ai Nguyen, Alan Herron, Lawrence A Donehower
ZUSAMMENFASSUNG
Mutation of K-Ras is a frequent oncogenic event in human cancers, particularly cancers of lungs, pancreas, and colon. It remains unclear why some tissues are more susceptible to Ras-induced transformation than others. Here, we globally activated a mutant oncogenic K-Ras allele (K-Ras(G12D)) in mice and examined the tissue-specific effects of this activation on cancer pathobiology, Ras signaling, tumor suppressor, DNA damage, and inflammatory responses. Within 5 to 6 weeks of oncogenic Ras activation, mice develop oral and gastric papillomas, lung adenomas, and hematopoietic hyperproliferation and turn moribund. The oral, gastric, and lung premalignant lesions display activated extracellular signal-regulated kinases (Erk)1/2 and NF-κB signaling as well as activated tumor suppressor and DNA damage responses. Other organs such as pancreas, liver, and small intestine do not exhibit neoplastic progression within 6 weeks following K-Ras(G12D) activation and do not show a potent tumor suppressor response. Even though robust Erk1/2 signaling is activated in all the tissues examined, the pErk1/2 distribution remains largely cytoplasmic in K-Ras(G12D)-refractory tissues (pancreas, liver, and intestines) as opposed to a predominantly nuclear localization in K-Ras(G12D)-induced neoplasms of lung, oral, and gastric mucosa. The downstream targets of Ras signaling, pElk-1 and c-Myc, are elevated in K-Ras(G12D)-induced neoplastic lesions but not in K-Ras(G12D)-refractory tissues. We propose that oncogenic K-Ras-refractory tissues delay oncogenic progression by spatially limiting the efficacy of Ras/Raf/Erk1/2 signaling, whereas K-Ras-responsive tissues exhibit activated Ras/Raf/Erk1/2 signaling, rapidly form premalignant tumors, and activate potent antitumor responses that effectively prevent further malignant progression.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-phospho-Histon H2A.X (Ser139)-Antikörper, Klon JBW301, clone JBW301, Upstate®, from mouse
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
ChIPAb+ NFκB p65 (RelA) - durch ChIP validiertes Antikörper- und Primer-Set, from mouse
Preise und Verfügbarkeit sind derzeit nicht verfügbar.