Merck
  • Asymmetric dimethylarginine induces apoptosis via p38 MAPK/caspase-3-dependent signaling pathway in endothelial cells.
Asymmetric dimethylarginine induces apoptosis via p38 MAPK/caspase-3-dependent signaling pathway in endothelial cells.
Journal of molecular and cellular cardiology (2006-03-07)
De-Jian Jiang, Su-Jie Jia, Zhong Dai, Yuan-Jian Li
ZUSAMMENFASSUNG
Asymmetric dimethylarginine (ADMA), an endogenous inhibitor of nitric oxide synthase (NOS), is emerging as a key contributor for endothelial dysfunction and its effects on endothelium are not yet completely defined. The aim of this study was to investigate ADMA-induced apoptosis and its mechanisms in human umbilical vein endothelial cells (HUVECs). Apoptosis was evaluated by in situ terminal uridine nick end labeling (TUNEL) assay and DNA fragmentation analysis. Caspase-3 activity was measured using a colorimetric protease assay kit. Activations of mitogen-activated protein kinases (MAPKs) were characterized by Western blot and immunofluorescence. Intracellular oxidant production was measured using H(2)DCF-DA, an oxidant-sensitive fluorescent indicator. ADMA (3-30 microM) induced apoptosis of HUVECs in a dose- and time-dependent manner. Caspase-3 was activated during apoptosis and its specific inhibitor DEVD-CHO significantly attenuated ADMA-induced apoptosis. Phosphorylation of p38 MAPK was induced by ADMA, and p38 MAPK specific inhibitor SB203580 concentration-dependently prevented ADMA-induced caspase-3 activation and cell apoptosis. ADMA increased intracellular oxidant production, which was significantly suppressed by intracellular antioxidant PDTC, l-arginine or antisense endothelial NOS mRNA. They also markedly prevented ADMA-induced phosphorylation of p38 MAPK and cell apoptosis. In conclusion, our present results demonstrate that ADMA induces apoptosis of endothelial cell via elevation of intracellular oxidant production, which involves p38 MAPK/caspase-3-dependent signaling pathway.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Caspase 3 Assay Kit, Colorimetric
Preise und Verfügbarkeit sind derzeit nicht verfügbar.