Merck
Rat colitis induced by intrathecal injection of substance P.
Sheng li xue bao : [Acta physiologica Sinica] (2009-08-25)
Ping Lin, Xing-Yu Wu, Hui Pan, Hui-Jun Jiang, Lin Mei
ZUSAMMENFASSUNG
The aim of this study was to, from the point of neurogenic inflammation, explore the pathogenesis of colitis and to provide direct evidence for the neurogenic colitis hypothesis. Male Sprague-Dawley rats (180-220 g) anesthetized with chloral hydrate were intrathecally (ith) implanted with polyethylene-10 (PE-10) catheter to reach the spinal cord T₁₂-L₅ level. Substance P (SP) was ith injected once a day for 14 d. The disease active index (DAI) score was calculated by rat body weight and stool. The macroscopic and HE staining-microscopic pathologies of colon/spinal tissue were evaluated. By immunofluorescence staining, the protein expression of a pro-inflammatory cytokine, migration inhibitory factor (MIF), in colon tissue was detected and was semi-quantitatively analyzed. The results showed that in the colon tissue, inflammation was dose-dependently aggravated by ith SP 10 μ and 20 μ, whereas in the spinal tissue, only slight edema and congestion were seen in SP 20 μ group. The MIF protein of colon tissue was increased in ith SP 10 μ and 20 μ groups (P<0.05, P<0.01 as compared to normal saline group respectively), but in the spinal tissue, there was no obvious MIF protein expression either in SP groups or in normal saline group. Pretreatment with neurokinin-1 (NK₁) receptor antagonist ([D-Pro2, D-Trp7, 9] -SP, 22.4 μ, ith, 10 min before ith SP) prolonged the latency of DAI rising and reduced the DAI amplitude, as well as prevented the high MIF expression induced by ith SP. These results suggest that rat colitis can be induced by direct SP stimulation in lumbar spine via activating central NK₁ receptor; and that colonic MIF is possibly one of the inflammatory factors involved in this pathogenesis. These data provide a reasonable support to the hypothesis of colitis being a neurogenic inflammation. In addition, a potential clinical significance for the finding that higher concentration of spinal SP can induce colitis via NK₁ receptor is discussed.