Merck
  • Purification and identification of the fusicoccin binding protein from oat root plasma membrane.
Purification and identification of the fusicoccin binding protein from oat root plasma membrane.
Plant physiology (1989-01-01)
A H de Boer, B A Watson, R E Cleland
ZUSAMMENFASSUNG
Fusicoccin (FC), a fungal phytotoxin, stimulates the H(+) -ATPase located in the plasma membrane (PM) of higher plants. The first event in the reaction chain leading to enhanced H(+) -efflux seems to be the binding of FC to a FC-binding protein (FCBP) in the PM. We solubilized 90% of the FCBP from oat (Avena sativa L. cv Victory) root PM in an active form with 1% octyl-glucoside. The FCBP was stabilized by the presence of protease inhibitors. The FCBP was purified by affinity chromatography using FC-linked adipic acid dihydrazide agarose (FC-AADA). Upon elution with 8 molar urea, two major protein bands on sodium dodecyl sulfate-polyaerylamide gel electrophoresis with molecular weights of 29,700 and 31,000 were obtained. Successive chromatography on BBAB Bio-Gel A, hexyl agarose, and FC-AADA resulted in the same two bands when the FC-AADA was eluted with sodium dodecyl sulfate. A direct correlation was made between 3H-FC-binding activity and the presence of the two protein bands. The stoichiometry of the 29,700 and 31,000 molecular weight bands was 1:2. This suggests that the FCBP occurs in the native form as a heterotrimer with an apparent molecular weight of approximately 92,000.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Adipinsäuredihydrazid-Agarose, saline suspension
Preise und Verfügbarkeit sind derzeit nicht verfügbar.