Merck
  • Regulation of glutamine synthetase. X. Effect of growth conditions on the susceptibility of Escherichia coli glutamine synthetase to feedback inhibition.
Regulation of glutamine synthetase. X. Effect of growth conditions on the susceptibility of Escherichia coli glutamine synthetase to feedback inhibition.
Journal of bacteriology (1967-10-01)
H S Kingdon, E R Stadtman
ZUSAMMENFASSUNG
The kinetic properties of Escherichia coli glutamine synthetase are markedly influenced by the manner in which the organism is grown. Enzyme obtained from stationary-phase cells grown on glycerol and glutamate is strongely inhibited by each of the eight feedback effectors known to influence this enzyme; however, the enzyme from log-phase cells grown on glucose and growth-limiting concentrations of NH(4)Cl is stimulated by some of these effectors. Of the growth variables examined, nitrogen source and time of harvest were the most important; carbon source and aeration seemed to have no effect. Two purified enzyme preparations have been obtained from cells grown under two different conditions, designated enzymes I and II for convenience. Enzyme I is stimulated by adenosine 5'-monophosphate, histidine, and tryptophan in the transfer assay, whereas enzyme II is strongly inhibited by all effectors tested. Enzyme I has a higher specific activity in the forward assay in the presence of Mg(++) or Co(++), whereas enzyme II is more active in the presence of Mn(++).