- A class of ruthenium(II) catalyst for asymmetric transfer hydrogenations of ketones.
A class of ruthenium(II) catalyst for asymmetric transfer hydrogenations of ketones.
Journal of the American Chemical Society (2005-05-19)
Aidan M Hayes, David J Morris, Guy J Clarkson, Martin Wills
PMID15898773
ZUSAMMENFASSUNG
Ruthenium dimer 6 (readily available in two steps from TsDPEN) is converted directly to monomeric asymmetric transfer hydrogenation catalyst 3 in situ under the conditions employed for ketone reduction. Catalyst 3 is a significantly more active catalyst for this application than the untethered derivative, exhibits higher enantioselectivities across a range of substrates, and appears to be highly stable to the reaction conditions. It is active at loadings of as low as 0.01 mol %, and reductions at the 0.1 mol % level are complete within 20 min at 80 degrees C without significant loss of enantioselectivity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Preise und Verfügbarkeit sind derzeit nicht verfügbar.