Merck
  • Definition of a FoxA1 Cistrome that is crucial for G1 to S-phase cell-cycle transit in castration-resistant prostate cancer.
Definition of a FoxA1 Cistrome that is crucial for G1 to S-phase cell-cycle transit in castration-resistant prostate cancer.
Cancer research (2011-09-09)
Chunpeng Zhang, Liguo Wang, Dayong Wu, Hongyan Chen, Zhong Chen, Jennifer M Thomas-Ahner, Debra L Zynger, Jérôme Eeckhoute, Jindan Yu, Jun Luo, Myles Brown, Steven K Clinton, Kenneth P Nephew, Tim H-M Huang, Wei Li, Qianben Wang
ZUSAMMENFASSUNG
The enhancer pioneer transcription factor FoxA1 is a global mediator of steroid receptor (SR) action in hormone-dependent cancers. In castration-resistant prostate cancer (CRPC), FoxA1 acts as an androgen receptor cofactor to drive G₂ to M-phase cell-cycle transit. Here, we describe a mechanistically distinct SR-independent role for FoxA1 in driving G₁ to S-phase cell-cycle transit in CRPC. By comparing FoxA1 binding sites in prostate cancer cell genomes, we defined a codependent set of FoxA1-MYBL2 and FoxA1-CREB1 binding sites within the regulatory regions of the Cyclin E2 and E2F1 genes that are critical for CRPC growth. Binding at these sites upregulate the Cyclin E2 and Cyclin A2 genes in CRPC but not in earlier stage androgen-dependent prostate cancer, establishing a stage-specific role for this pathway in CRPC growth. Mechanistic investigations indicated that FoxA1, MYBL2, or CREB1 induction of histone H3 acetylation facilitated nucleosome disruption as the basis for codependent transcriptional activation and G₁ to S-phase cell-cycle transit. Our findings establish FoxA1 as a pivotal driver of the cell-cycle in CRPC which promotes G₁ to S-phase transit as well as G₂ to M-phase transit through two distinct mechanisms.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
ChIPAb+ CREB - ChIP Validated Antibody and Primer Set, rabbit monoclonal, culture supernatant, clone NL904, from rabbit
Preise und Verfügbarkeit sind derzeit nicht verfügbar.