Merck
  • Ectopic adenine nucleotide translocase activity controls extracellular ADP levels and regulates the F1-ATPase-mediated HDL endocytosis pathway on hepatocytes
Ectopic adenine nucleotide translocase activity controls extracellular ADP levels and regulates the F1-ATPase-mediated HDL endocytosis pathway on hepatocytes
Biochimica et Biophysica Acta - Molecular and Cell Biology of Lipids (2017)
Cardouat G, et al.