Merck
  • Resistance of postharvest biocontrol yeasts to oxidative stress: a possible new mechanism of action
Resistance of postharvest biocontrol yeasts to oxidative stress: a possible new mechanism of action
Phytopathology (2003)
Castoria R, et al.