Merck
  • Is the effect of precipitation on acute gastrointestinal illness in southwestern Uganda different between Indigenous and non-Indigenous communities?
Is the effect of precipitation on acute gastrointestinal illness in southwestern Uganda different between Indigenous and non-Indigenous communities?
PloS one (2019-05-03)
Johanna Busch, Lea Berrang-Ford, Sierra Clark, Kaitlin Patterson, Emma Windfeld, Blanaid Donnelly, Shuaib Lwasa, Didacus Namanya, Sherilee L Harper
ZUSAMMENFASSUNG
Acute gastrointestinal illness (AGI) is a global public health priority that often disproportionately effects Indigenous populations. While previous research examines the association between meteorological conditions and AGI, little is known about how socio-cultural factors may modify this relationship. This present study seeks to address this research gap by comparing AGI prevalence and determinants between an Indigenous and non-Indigenous population in Uganda. We estimate the 14-day self-reported prevalence of AGI among adults in an Indigenous Batwa population and their non-Indigenous neighbours using cross-sectional panel data collected over four periods spanning typically rainy and dry seasons (January 2013 to April 2014). The independent associations between Indigenous status, precipitation, and AGI are examined with multivariable multi-level logistic regression models, controlling for relative wealth status and clustering at the community level. Estimated prevalence of AGI among the Indigenous Batwa was greater than among the non-Indigenous Bakiga. Our models indicate that both Indigenous identity and decreased levels of precipitation in the weeks preceding the survey period were significantly associated with increased AGI, after adjusting for confounders. Multivariable models stratified by Indigenous identity suggest that Indigenous identity may not modify the association between precipitation and AGI in this context. Our results suggest that short-term changes in precipitation affect both Indigenous and non-Indigenous populations similarly, though from different baseline AGI prevalences, maintaining rather than exacerbating this socially patterned health disparity. In the context of climate change, these results may challenge the assumption that changing weather patterns will necessarily exacerbate existing socially patterned health disparities.