Merck
  • Bioactive composition and sensory evaluation of innovative spaghetti supplemented with free or α-cyclodextrin chlatrated pumpkin oil extracted by supercritical CO2.
Bioactive composition and sensory evaluation of innovative spaghetti supplemented with free or α-cyclodextrin chlatrated pumpkin oil extracted by supercritical CO2.
Food chemistry (2019-05-28)
Miriana Durante, Marcello S Lenucci, Laura Gazza, Federica Taddei, Francesca Nocente, Giuseppe E De Benedetto, Monica De Caroli, Gabriella Piro, Giovanni Mita
ZUSAMMENFASSUNG
The feasibility of producing durum wheat pasta enriched with a lipophilic phytocomplex, extracted using supercritical carbon dioxide (SC-CO2), from ripe pumpkin, as free oil or as ready-to-mix oil/α-cyclodextrins (α-CDs) powder, was explored. Four types of pasta were prepared: (i) control spaghetti (S-CTRL); (ii) spaghetti supplemented with α-CDs (S-α-CD); (iii) spaghetti supplemented with pumpkin oil (S-Oil) and (iv) spaghetti supplemented with the pumpkin oil/α-CD powder (S-Oil/α-CD). The chemical, antioxidant, textural and sensory attributes of the different pasta were evaluated and compared. S-Oil and S-Oil/α-CD spaghetti were significantly enriched with phytosterols, squalene, carotenoids, tocochromanols and unsaturated fatty acids. Spaghetti containing α-CDs were slightly improved in terms of fiber content. Oil chlatration increased the stability of some bioactives during pasta production and ameliorated poor textural and sensory characteristics of the cooked spaghetti compared with S-Oil sample. S-Oil/α-CD spaghetti might be accepted by customers, if the potential health benefits were also explained.