Merck
  • Towards a solution to MERS: protective human monoclonal antibodies targeting different domains and functions of the MERS-coronavirus spike glycoprotein.
Towards a solution to MERS: protective human monoclonal antibodies targeting different domains and functions of the MERS-coronavirus spike glycoprotein.
Emerging microbes & infections (2019-04-03)
Ivy Widjaja, Chunyan Wang, Rien van Haperen, Javier Gutiérrez-Álvarez, Brenda van Dieren, Nisreen M A Okba, V Stalin Raj, Wentao Li, Raul Fernandez-Delgado, Frank Grosveld, Frank J M van Kuppeveld, Bart L Haagmans, Luis Enjuanes, Dubravka Drabek, Berend-Jan Bosch
ZUSAMMENFASSUNG
The Middle-East respiratory syndrome coronavirus (MERS-CoV) is a zoonotic virus that causes severe and often fatal respiratory disease in humans. Efforts to develop antibody-based therapies have focused on neutralizing antibodies that target the receptor binding domain of the viral spike protein thereby blocking receptor binding. Here, we developed a set of human monoclonal antibodies that target functionally distinct domains of the MERS-CoV spike protein. These antibodies belong to six distinct epitope groups and interfere with the three critical entry functions of the MERS-CoV spike protein: sialic acid binding, receptor binding and membrane fusion. Passive immunization with potently as well as with poorly neutralizing antibodies protected mice from lethal MERS-CoV challenge. Collectively, these antibodies offer new ways to gain humoral protection in humans against the emerging MERS-CoV by targeting different spike protein epitopes and functions.