Merck
  • A facile approach for engineering tissue constructs with vessel-like channels by cell-laden hydrogel fibers.
A facile approach for engineering tissue constructs with vessel-like channels by cell-laden hydrogel fibers.
Materials science & engineering. C, Materials for biological applications (2019-04-29)
Meiling Zhong, Xiaolu Liu, Dan Wei, Jing Sun, Likun Guo, Hua Zhu, Yizao Wan, Hongsong Fan
ZUSAMMENFASSUNG
Vascularization is of great importance in the successful translation of tissue engineered constructs into clinically relevant application. The lack of a general approach to rapidly construct vascular networks in engineered constructs remains a major challenge. Herein, an adhesive hydrogel-based tissue construct, in which cell-affinitive domains and interconnected channels were concurrently constructed, was put forward to enhance vascularization. Hydrogel matrix was modified with Arg-Gly-Asp (RGD) peptide to supply cell adhesion sites. Collagen fibers were added into the hydrogel matrix to produce interconnected vessel-like channels via enzyme mediated degradation. In a bone-like model, the successful outspread morphology and intensive function expression of osteo-like cells and the formation of endothelial cells-lined channels were observed, suggesting it's flexible to functionalize extracellular matrix with vessel-like channels via the introduction of endothelial cells-laden fibers. Our approach furnishes a particular strategy to build vascular architecture and is especially attractive in the bioengineering of rich vascularized tissues.