Merck
  • Long-term functional and structural preservation of precision-cut human myocardium under continuous electromechanical stimulation in vitro.
Long-term functional and structural preservation of precision-cut human myocardium under continuous electromechanical stimulation in vitro.
Nature communications (2019-01-12)
Carola Fischer, Hendrik Milting, Evelyn Fein, Elisabeth Reiser, Kun Lu, Thomas Seidel, Camilla Schinner, Thomas Schwarzmayr, Rene Schramm, Roland Tomasi, Britta Husse, Xiaochun Cao-Ehlker, Ulrich Pohl, Andreas Dendorfer
ZUSAMMENFASSUNG
In vitro models incorporating the complexity and function of adult human tissues are highly desired for translational research. Whilst vital slices of human myocardium approach these demands, their rapid degeneration in tissue culture precludes long-term experimentation. Here, we report preservation of structure and performance of human myocardium under conditions of physiological preload, compliance, and continuous excitation. In biomimetic culture, tissue slices prepared from explanted failing human hearts attain a stable state of contractility that can be monitored for up to 4 months or 2000000 beats in vitro. Cultured myocardium undergoes particular alterations in biomechanics, structure, and mRNA expression. The suitability of the model for drug safety evaluation is exemplified by repeated assessment of refractory period that permits sensitive analysis of repolarization impairment induced by the multimodal hERG-inhibitor pentamidine. Biomimetic tissue culture will provide new opportunities to study drug targets, gene functions, and cellular plasticity in adult human myocardium.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonales Anti-α-Aktinin (Sarkomer) in Maus hergestellte Antikörper, clone EA-53, ascites fluid
Sigma-Aldrich
Anti--α-Glattmuskel-Aktin-Antikörper, Maus, monoklonal, clone 1A4, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Connexin-43 in Kaninchen hergestellte Antikörper, affinity isolated antibody, buffered aqueous solution