Merck
  • Classical swine fever virus non-structural proteins modulate Toll-like receptor signaling pathways in porcine monocyte-derived macrophages.
Classical swine fever virus non-structural proteins modulate Toll-like receptor signaling pathways in porcine monocyte-derived macrophages.
Veterinary microbiology (2019-03-05)
Zhi Cao, Qian Yang, Minping Zheng, Huifang Lv, Kai Kang, Yanming Zhang
ZUSAMMENFASSUNG
Toll-like receptors (TLRs) are crucial activators of the innate immune response that play various roles in viral infection. Studies have confirmed that classical swine fever virus (CSFV) infection has significant effects on the expression of immune effectors participating in TLR signaling pathways; however, the involvement of CSFV-encoded proteins in TLR signaling pathways remains unclear. In this study, lentiviral individually expressing CSFV non-structural proteins (NSPs) were constructed to identify the "key proteins" that affect TLR gene expression and to analyze the impacts of these proteins on factors downstream of the TLR signaling pathways. The results indicated that Npro, NS2, NS3, NS3/4A, NS4B and NS5A all failed to induce the activation of NF-κB p65. Furthermore, NS4B was found to inhibit poly (I:C) stimulation-mediated activation of the TLR3 signaling pathway in porcine monocyte-derived macrophages (pMDMs), thereby suppressing the TRIF mRNA transcription, the IRF3 protein translation and the NF-κB p65 phosphorylation, and ultimately affecting the secretion of IL-6 and IFN-β; CSFV NS5A protein could significantly increase the activation of MyD88 and IRF7 as well as the consequent synthesis of IFN-α in pMDMs. The results suggest that CSFV NSPs affect TLR-mediated innate immune responses in pMDMs.